Select a Size
About This Item
Product Name
Anti-Cas9 Antibody, C-terminal Antibody, clone 5F1 Antibody, S. aureus, clone 5F1, from mouse
biological source
mouse
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
5F1, monoclonal
species reactivity
Staphylococcus aureus, bacteria
species reactivity (predicted by homology)
all
packaging
antibody small pack of 25 μg
technique(s)
western blot: suitable
isotype
IgG1κ
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Quality Level
Analysis Note
Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .
Application
Secondary & Control Antibodies
Biochem/physiol Actions
Disclaimer
General description
Immunogen
Other Notes
Physical form
Preparation Note
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service