MABX31
Anti-Zika Virus (ZIKV-NS1)
Synonym(s):
Anti-ZIKV-NS1 Antibody
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
clone
monoclonal
form
solution
species reactivity
all
packaging
pkg of 1 mg (MABX31-1KC)
pkg of 10 mg (MABX31-10KC)
technique(s)
ELISA: suitable
immunodiffusion: suitable
isotype
IgG1
relevant disease(s)
infectious diseases
shipped in
wet ice
storage temp.
2-8°C
Specificity
No reactivity with Dengue Virus or Chikungunya Virus
Immunogen
Recombinant Zika Virus NS1 Protein
Application
This mouse monoclonal Anti-Zika Virus (ZIKV-NS1), Cat. No. MABX31, is tested for use in Lateral Flow Assay and Enzyme Immunoassay (ELISA), for the detection of Recombinant Zika Virus NS1 Protein.
Target description
Zika virus (ZIKV) is a member of the virus family Flaviviridae. Zika is spread mostly by the bite of infected Aedes species mosquitoes, such as Ae. aegypti and Ae. albopictus. Its name comes from the Ziika Forest of Uganda, where the virus was first isolated in 1947. Zika can be passed from a pregnant woman to her fetus. Infection during pregnancy can cause certain birth defects.
Linkage
Recommended Matched Pair MABX30 and MABX31
Physical form
Purified mouse monoclonal in buffer containing 0.015 M Potassium Phosphate, pH 7.2 with 0.85% Sodium Chloride and 0.05% Sodium Azide
Reconstitution
Please refer to Certificate of Analysis for concentration.
Storage and Stability
Store at 2-8 °C. For long-term storage, freeze at -20 °C
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service