Skip to Content
Merck
All Photos(2)

Key Documents

MABS466

Sigma-Aldrich

Anti-Trpv4 Antibody, clone 1B2.6

clone 1B2.6, from mouse

Synonym(s):

Trpv4, Transient receptor potential cation channel subfamily V member 4, TrpV4, Osm-9-like TRP channel 4, OTRPC4, Transient receptor potential protein 12, TRP12, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, VRL-2, Vanilloid recep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B2.6, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRPV4(59341)

General description

Transient receptor potential cation channel subfamily V member 4 is a multitransmembrane non-selective cation channel encoded by the TRPV4 (also known as CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VR-OAC, VRL-2, VRL2, VROAC) gene and belong to the OSM9-like transient receptor potential channel (OTRPC) subfamily within the transient receptor potential (TRP)3 superfamily of channels. Ankyrin (ANK) repeats/domains are shown to be important for functional channel expression, and ANK domain mutations are found in patients suffering from Spinal muscular atrophies (SMA, also known as hereditary motor neuropathies) and hereditary motor and sensory neuropathies (HMSN). (PMID 16293632, 20037587, 20037588).

Specificity

This antibody is expected to react to all isoforms.

Immunogen

GST-tagged recombinant protein corresponding to human Trpv4.

Application

Anti-Trpv4 Antibody, clone 1B2.6 is an antibody against Trpv4 for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected Trpv4 in NIH/3T3 cells.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in SK-MEL-28 cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected Trpv4 in 10 µg of SK-MEL-28 cell lysate.

Target description

~86-98 kDa observed. Uniprot describes 6 isoforms produced by alternative splicing ranging from ~86 to 98 kDa Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parto Babaniamansour et al.
Biomedicines, 12(1) (2024-01-26)
Investigating macrophage plasticity emerges as a promising strategy for promoting tissue regeneration and can be exploited by regulating the transient receptor potential vanilloid 4 (TRPV4) channel. The TRPV4 channel responds to various stimuli including mechanical, chemical, and selective pharmacological compounds.
NADPH oxidase 4 contributes to TRPV4-mediated endothelium-dependent vasodilation in human arterioles by regulating protein phosphorylation of TRPV4 channels.
Xie, et al.
Basic Research in Cardiology, 117, 24-24 (2023)
Kaustav Bera et al.
Nature, 611(7935), 365-373 (2022-11-04)
Cells respond to physical stimuli, such as stiffness1, fluid shear stress2 and hydraulic pressure3,4. Extracellular fluid viscosity is a key physical cue that varies under physiological and pathological conditions, such as cancer5. However, its influence on cancer biology and the mechanism
Charles E MacKay et al.
eLife, 9 (2020-05-05)
PKD2 (polycystin-2, TRPP1), a TRP polycystin channel, is expressed in endothelial cells (ECs), but its physiological functions in this cell type are unclear. Here, we generated inducible, EC-specific Pkd2 knockout mice to examine vascular functions of PKD2. Data show that
Nicole R Wong et al.
Immunity, 54(9), 2072-2088 (2021-07-29)
Cardiac macrophages represent a heterogeneous cell population with distinct origins, dynamics, and functions. Recent studies have revealed that C-C Chemokine Receptor 2 positive (CCR2+) macrophages derived from infiltrating monocytes regulate myocardial inflammation and heart failure pathogenesis. Comparatively little is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service