MABF2079M
Anti-CD8 alpha Antibody, clone 53-6.72
clone 53-6.72, from rat
Synonym(s):
T-cell surface glycoprotein CD8 alpha chain, T-cell surface glycoprotein Lyt-2
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
53-6.72, monoclonal
species reactivity
mouse
packaging
antibody small pack of 25 μg
technique(s)
flow cytometry: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
mouse ... Cd8A(12525)
Related Categories
General description
Specificity
Immunogen
Application
Functional Analysis: A representative lot detected CD8 alpha in Functional analysis applications (Daley, D., et. al. (2016). Cell. 166(6):1485-1499; Hannani, D., et. al. (2014). Cell Death Differ. 21(1):50-8).
Quality
Flow Cytometry Analysis: 1 µg of this antibody detected CD8 alpha in one million mouse thymocytes.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service