Skip to Content
Merck

MABF1968

Anti-Microglobulin beta2 Antibody, clone BBM.1

clone BBM.1, from mouse

Synonym(s):

B2M

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
BBM.1, monoclonal
Application:
FACS, IHC, WB, inhibition assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BBM.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable (paraffin), inhibition assay: suitable, western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... B2M(567)

General description

Beta-2-microglobulin (UniProt: P61769) is encoded by the B2M (also known as CDABP0092, HDCMA22P) gene (Gene ID: 567) in human. Beta-2-microglobulin is a component of the class I major histocompatibility complex (MHC) that is involved in the presentation of peptide antigens to the immune system. It is synthesized with a signal peptide of 20 amino acids, which is subsequently cleaved to generate the mature form. It also contains an Ig-like C1 type domain (aa 25-113). Beta-2-microglobulin lacks a transmembrane anchoring domain and is found in secreted as well as in complex. Serum beta-2-microglobulin levels are a reflection of cell turnover and its levels are shown to be elevated in fever, inflammation, and infection. Increased serum levels are also seen in B-cell malignancies and in renal failure and may indicate a worse prognosis for patients with early-stage Hodgkin′s lymphoma. Its levels may be elevated either due to higher rates of synthesis as observed in HIV infection and malignancy or due to impaired renal filtration due to renal insufficiency.
~14 kDa observed; 13.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

MOLT-4, a Human acute lymphoblastic leukemia cell line.

Application

Anti-Microglobulin beta2, clone BBM.1, Cat. No. MABF1968, is a mouse monoclonal antibody that detects Microglobulin beta2 and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), Inhibition assays, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Microglobulin beta2 in Immunohistochemistry applications (Josson, S., et. al. (2011). Cancer Res. 71(7):2600-10).

Inhibition Analysis: A representative lot inhibited Microglobulin beta2 activity (Parham, P., et. al. (1983). J Biol Chem. 258(10):6179-86).

Flow Cytometry Analysis: A representative lot detected Microglobulin beta2 in Flow Cytometry applications (Drukker, M., et. al. (2012). Nat Biotechnol. 30(6):531-42).

Immunohistochemistry Analysis: A 1:50-1:1,000 dilution from a representative lot detected Microglobulin beta2 in human breast cancer and human tonsil tissues.
Research Category
Inflammation & Immunology

Biochem/physiol Actions

Clone BBM.1 detects Beta-2-microglobulin in human.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in MOLT-4 cell lysates

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Microglobulin beta2 in 10 µg of MOLT-4 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library