Skip to Content
Merck

MABE951

Anti-Histone H3.3 (H3-3A,H3-3B) Antibody

rat monoclonal, 6C4A3

Synonym(s):

Histone H3.3, H3.3A, H3F3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
6C4A3, monoclonal
Species reactivity:
human, monkey, mouse, canine
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Histone H3.3 Antibody, clone 6C4A3, clone 6C4A3, 1 mg/mL, from rat

biological source

rat

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C4A3, monoclonal

species reactivity

human, monkey, mouse, canine

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
~17 kDa observed

Application

Anti-Histone H3.3 Antibody, clone 6C4A3 is an antibody against Histone H3.3 for use in western blotting.
Western Blotting Analysis: 0.1 µg/mL from a representative lot detected Histone H3.3 with recombinant H3.3 proteins. Additionally, 1 µg/mL from a representative lot detected Histone H3.3 in HeLa, COS1, NIH/3T3, NRK, and MDCK whole cell lysates (Prof. Taro Tachibana, Cell Engineering Corporation.).

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: 1 µg/mL of this antibody detected Histone H3.3 in 10 µg of HeLa acid extract.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library