Skip to Content
Merck

MABE283

Anti-p53 Antibody, clone PAb421

clone PAb421, from mouse

Synonym(s):

Cellular tumor antigen p53, Tumor suppressor p53

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
PAb421, monoclonal
Application:
IHC, IP, WB
Citations:
39
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PAb421, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Trp53(22059)

General description

p53 was discovered in 1979 as a cellular protein associating with the transforming protein of SV40 tumor virus. Since then, many different biochemical functions have been attributed to the 53 kDa phosphoprotein. Experimental evidence has suggested that p53 acts as a negative regulator of cell growth in normal cells). Thus, the inactivation or mutation of p53 may be an essential step in the development of malignancy. Wild-type p53 levels in normal cells and tissues were found to be very low. Mutant p53 polypeptide, however, is often found to be present at high concentrations in mammalian tumors and tumor cell lines. For example, in an immuno-histochemistry study 40% of human breast cancer showed elevated levels of mutant p53 in the cell nucleus. Mutations of the p53 protein have some characteristic features: Most of them are missense point mutations giving rise to an altered protein function. Also, many, but not all, mutant p53 proteins exhibit a common mutant structure, which can be recognized by monoclonal antibodies specific for p53 in the mutant conformation.
~53 kDa observed

Immunogen

Epitope: Amino acids 376-378 in human p53
Partially purified mouse p53

Application

Immunoprecipitation Analysis: A representative lot from independent laboratory immunoprecipitated p53 in IP (Lehman, T. A., et al. (1991). Cancer Res. 51(15):4090-4096.; Harlow, E., et al. (1981). J Virol. 39(3):861-869.)

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected p53 in human breast cancer tissue (Davidoff, A. M., et al. (1992). Proc Natl Acad Sci USA. 89(8):3439-3442.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-p53 Antibody, clone PAb421 (Mouse Monoclonal Antibody) validated in WB, IP, IHC to detect p53 also known as Cellular tumor antigen p53, Tumor suppressor p53.

Biochem/physiol Actions

This antibody recognizes amino acids 376-378 of human p53.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Mouse brain tissue lysate
Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected p53 in 10 µg of mouse brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins






Global Trade Item Number

SKUGTIN
MABE28304053252646317