Skip to Content
Merck

MABC674

Anti-LPL Antibody, clone 2C5

ascites fluid, clone 2C5, from mouse

Synonym(s):

Lipoprotein lipase, HDLCQ11

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2C5, monoclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2C5, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LPL(4023)

General description

Lipoprotein lipase (LPL) is encoded by the LPL/LIPD gene in humans and is a lipase that localizes to the plasma membrane of the microvilli of hepatocytes and the luminal surface of endothelial cells in capillaries. It is expressed in a wide variety of tissues from fat and heart to muscle and mammary glands. It can also be found in the blood bound to lipoproteins. Its function is to hydrolyze triglycerides from the blood and other sources into free fatty acids; LPL also acts as receptor and binding factor for lipoprotein uptake and allows for the catalysis of VLDL from the blood. Expression and regulation of LPL differ by tissue type, some factors such as insulin can stimulate LPL expression in fat tissues, where as insulin down regulates LPL expression on muscle. Recently because serum triglyceride levels are linked with the development of cancer, LPL has become a target for cancer prevention and therapy because LPL catalyzes plasma triglycerides leading to lower levels, it is thought that LPL assisted therapy could be useful in preventing or treating cancers. EMD-Millipore’s Anti-LPL mouse monoclonal antibody has been tested on western blots using HeLa cell lysates.
~53 kDa observed

Immunogen

Purified recombinant fragment of human LPL expressed in E. Coli.

Application

Anti-LPL Antibody, clone 2C5 is a mouse monoclonal antibody & validated for use in western blotting.

Analysis Note

Control
Hela cell lysate
Evaluated by Western Blotting in Hela cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected LPL in Hela cell lysate.

Optimal working dilutions must be determined by end user.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library