Skip to Content
Merck

MABC573

Anti-Bcl-2 Antibody, clone 1D7.2

clone 1D7.2, from rat

Synonym(s):

Apoptosis regulator Bcl-2, Bcl-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1D7.2, monoclonal
Application:
WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D7.2, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Bcl2(12043)

General description

Apoptosis regulator Bcl-2 (UniProt P10417; also known as B-cell leukemia/lymphoma 2) is encoded by the Bcl2 (also known as Bcl-2, AW986256, C430015F12Rik, D630044D05Rik, D830018M01Rik) gene (Gene ID 12043) in murine species. Bcl-2 is an anti-apoptotic and the founding member of the Bcl-2 family of proteins that paly either a pro- or anti-apoptotic role in cell death signaling. BCL-2 family members are related by regions of sequence and structural homology. The pro-survival members contain four BCL-2 homology regions (BH1–BH4), whereas the BH3-only members contain only the BH3 amphipathic helix, which mediates their interaction with the groove of multi-domain BCL-2 family members. Human BCL2 gene mutatons are well known causes of many cancers, including melanoma, breast, prostate, chronic lymphocytic leukemia, and lung cancer.
~22 kDa observed (26.4 kDa/isoform 1 & 22.3 kDa/isoform 2, calculated).

Immunogen

Epitope: N-terminal half
GST-tagged recombinant protein corresponding to the N-terminal of mouse Bcl-2.

Application

Detect Bcl-2 using this rat Monoclonal Anti-Bcl-2 Antibody, clone 1D7.2, Cat. No. MABC573, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

Expected to react with both spliced isoforms of murine Bcl-2.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse myoblast C2C12 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Bcl-2 in 10 µg of mouse myoblast C2C12 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

A major focus of breast cancer research is to understand the mechanisms responsible for disease progression and drug resistance. Toward that end, it has been found that approximately two thirds of all human breast carcinomas overexpress the Estrogen Receptor α (ERα) protein and it remains the primary pharmacological target for endocrine therapy1,2. The normal cellular function of ERα is as a transcription factor that mediates a wide variety of physiological processes, many of which are dependent upon phosphorylation of the receptor at specific amino acid residues3,4. Indeed, ERα is known to be phosphorylated at a multitude of different sites, yet how these all correlate to disease remains unclear5. Here, we interrogated multiple sites of ERα for phosphorylation status by screening an extensive panel of different breast cancer patient samples and other non-breast cancer tissue microarray (TMA) slide samples to determine their relevance to disease.






Global Trade Item Number

SKUGTIN
MABC57304055977176384