Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
CV5.1, monoclonal
Application:
ICC
Citations:
4
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
CV5.1, monoclonal
species reactivity
human
concentration
0.5 mg/mL
technique(s)
immunocytochemistry: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FANCM(57697)
General description
230 kDa calculated
FANCM, also known as Fanconi anemia group M protein (FACM), or ATP-dependent RNA helicase FANCM, or Fanconi anemia-associated polypeptide of 250 kDa (FAAP250), or Protein Hef ortholog, and encoded by the gene FANCM/KIAA1596,is a critical ATPase needed for proper DNA repair. FANCM is found at DNA replication forks and required for cellular resistance to DNA cross-linking agents and for proper DNA repair. FANCM belongs to a multi-subunit FA (Fanconi anemia) complex. FANCM is localized to the nucleus and expressed in most cells. Defects in FANCM may be associated with Fanconi anemia complementation group M (FANCM) a disorder affecting hematopoietic cells can causing anemia, leukopenia and thrombopenia. Patients also have cardiac, renal and limb malformations, dermal pigmentary changes, and a predisposition toward malignancies. Patient’s cells show hypersensitivity to DNA-damaging agents, chromosomal instability and defective DNA repair.
Immunogen
His-tagged recombinant protein corresponding to human FANCM.
Application
Detect FANCM using this Anti-FANCM Antibody, clone CV5.1 validated for use in ICC.
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
Physical form
Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Immunohistochemistry Analysis in A431, HeLa, and HepG2 cells.
Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected FANCM in A431, HeLa, and HepG2 cells.
Dylight® is a registered trademark of Thermo Fisher Scientific.
Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected FANCM in A431, HeLa, and HepG2 cells.
Dylight® is a registered trademark of Thermo Fisher Scientific.
Legal Information
DyLight is a registered trademark of Pierce Biotechnology, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Massimo Bogliolo et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 20(4), 458-463 (2017-08-25)
PurposeMutations in genes involved in Fanconi anemia (FA)/BRCA DNA repair pathway cause cancer susceptibility diseases including familial breast cancer and Fanconi anemia (FA). A single FA patient with biallelic FANCM mutations was reported in 2005 but concurrent FANCA pathogenic mutations
Gisella Figlioli et al.
NPJ breast cancer, 5, 38-38 (2019-11-09)
Breast cancer is a common disease partially caused by genetic risk factors. Germline pathogenic variants in DNA repair genes BRCA1, BRCA2, PALB2, ATM, and CHEK2 are associated with breast cancer risk. FANCM, which encodes for a DNA translocase, has been
Chia-Yu Guh et al.
Nature communications, 13(1), 5781-5781 (2022-10-03)
Alternative Lengthening of Telomeres (ALT) utilizes a recombination mechanism and break-induced DNA synthesis to maintain telomere length without telomerase, but it is unclear how cells initiate ALT. TERRA, telomeric repeat-containing RNA, forms RNA:DNA hybrids (R-loops) at ALT telomeres. We show
Neus P Lorite et al.
Cellular and molecular life sciences : CMLS, 81(1), 251-251 (2024-06-07)
The Smc5/6 complex is a highly conserved molecular machine involved in the maintenance of genome integrity. While its functions largely depend on restraining the fork remodeling activity of Mph1 in yeast, the presence of an analogous Smc5/6-FANCM regulation in humans
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MABC545 | 04053252976315 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service