Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
11E12, monoclonal
Application:
IP, WB
Citations:
1
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
11E12, monoclonal
species reactivity
human, mouse
technique(s)
immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... VHL(7428)
General description
von Hippel-Lindau protein (VHL) is most widely recognized as belonging to an E3 ubiquitin ligase complex that specifically targets hypoxia-inducible factors (HIFs) for the purpose of polyubiquitination and proteolytic degradation. Within this complex, VHL acts as a target recruitment subunit for HIFα and its subsequent ubiquitination. The complex also contains proteins such as B and C elongins, Rbx1 and Cul2. Inactivation of VHL has been linked to renal cell carcinoma development, therefore suggesting that VHL plays a role as a tumor suppressor. VHL expression has been observed in both fetal and adult kidney and brain tissues. Defects in VHL expression are causal to renal cell carcinoma type 1 (RCC1), erythrocytosis familial type 2 (ECYT2), von Hippel-Lindau disease (VHLD), and pheochromocytoma.
~ 24 kDa observed MW. VHL SUMOylation may result in high MW bands (Cai, 2010).
Immunogen
Purified human VHL
Application
Immunoprecipitation: It has been shown that this VHL clone will immunoprecipitate VHL from HEK 293 cells (Shoenfeld, 1998).
Research Category
Apoptosis & Cancer
Signaling
Apoptosis & Cancer
Signaling
Research Sub Category
Tumor Markers
Ubiquitin & Ubiquitin Metabolism
Tumor Markers
Ubiquitin & Ubiquitin Metabolism
This Anti-VHL Antibody, clone 11E12 is validated for use in WB, IP for the detection of VHL.
Physical form
Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
Mouse fetal brain tissue lysate
Mouse fetal brain tissue lysate
Evaluated by Western Blot in mouse fetal brain tissue lysate.
Western Blot Analysis: 1 µg/mL of the antibody detected VHL in 10 µg of mouse fetal brain tissue lysate.
Western Blot Analysis: 1 µg/mL of the antibody detected VHL in 10 µg of mouse fetal brain tissue lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.