Skip to Content
Merck
All Photos(1)

Key Documents

MAB9410

Sigma-Aldrich

Anti-Coxsackievirus B Blend Antibody, clone Blend of 6s

ascites fluid, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
monoclonal
application:
IF
species reactivity:
human
technique(s):
immunofluorescence: suitable
citations:
6

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

General description

Blend of 6 monoclonals

Specificity

Reacts with the Coxsackie virus B1, B2, B3, B4, B5 and B6.

CROSS REACTIVITIES:

No cross-reactivity to CMV, HSV 1 & 2, Para 1, 2, &3, Poliovirus 1 & 3 or Mumps.

Application

Anti-Coxsackievirus B Blend Antibody, clone Blend of 6s is an antibody against Coxsackievirus B Blend for use in IF.
IFA at 1:100.

Dilute with buffer pH 7.5-8.0 containing protein or other stabilizing medium.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites.

Storage and Stability

Maintain at -20°C in convenient, undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuan-Chi Tseng et al.
Viruses, 14(1) (2022-01-23)
Enterovirus 71 (EV71) is an etiological agent of hand foot and mouth disease and can also cause neurological complications in young children. However, there are no approved drugs as of yet to treat EV71 infections. In this study, we conducted
Hsin-Yu Hou et al.
The Journal of general virology, 97(5), 1122-1133 (2016-02-18)
Enterovirus 71 (EV71) causes life-threatening diseases with neurological manifestations in young children. However, the treatment of EV71 infections remains an unmet medical need. Idarubicin (IDR) is an anthracycline compound that is used therapeutically for certain types of tumour. In this
Yu-Siang Su et al.
Frontiers in microbiology, 11, 1656-1656 (2020-08-08)
Enterovirus A71 (EV-A71) is one of the major etiologic agents causing hand, foot, and mouth disease (HFMD) in children and occasionally causes severe neurological diseases or even death. EV-A71 replicates rapidly in host cells. For a successful infection, viruses produce
Robert Root-Bernstein et al.
International journal of molecular sciences, 24(15) (2023-08-12)
Autoimmune cardiopathies (AC) following COVID-19 and vaccination against SARS-CoV-2 occur at significant rates but are of unknown etiology. This study investigated the possible roles of viral and bacterial mimicry, as well as viral-bacterial co-infections, as possible inducers of COVID-19 AC
Robert Root-Bernstein et al.
International journal of molecular sciences, 23(19) (2022-10-15)
COVID-19 patients often develop coagulopathies including microclotting, thrombotic strokes or thrombocytopenia. Autoantibodies are present against blood-related proteins including cardiolipin (CL), serum albumin (SA), platelet factor 4 (PF4), beta 2 glycoprotein 1 (β2GPI), phosphodiesterases (PDE), and coagulation factors such as Factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service