Skip to Content
Merck
All Photos(1)

Key Documents

MAB880-A

Sigma-Aldrich

Anti-HIV Antibody, p24, clone 7A8.1

clone 7A8.1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7A8.1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

-1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Gene Information

human ... TARBP1(6894)

Specificity

Core protein p24. No cross-reactivity observed to other HIV proteins.

Immunogen

Epitope: p24
p24 isolated from human (native) HIV infected cells.

Application

Anti-HIV Antibody, p24, clone 7A8.1 is an antibody against HIV for use in IC & WB.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Western Blot (1:100) or Immunoperoxidase or Immunofluorescence staining on human tissue (1:100).

Antibodies are not neutralizing. For extensive dilution, protein containing or other stabilizing medium should be used. Final working dilutions must be determined by end user.

Linkage

Replaces: MAB882

Physical form

Format: Purified
Purified in 0.02M phosphate buffer, pH7.6, 0.25M NaCl, containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Liu et al.
The Journal of infectious diseases, 208(8), 1221-1230 (2013-07-16)
Many chronic human immunodeficiency virus (HIV) patients suffer from gastric complaints, including gastric tuberculosis and coinfection of other pathogens. Recent work has demonstrated that a variety of nonimmune cells can act as viral reservoirs, even at the early stage of
Shilpi Sharma et al.
Journal of virology, 93(11) (2019-03-15)
The cellular protein bone marrow stromal antigen-2 (BST-2)/tetherin acts against a variety of enveloped viruses by restricting their release from the plasma membrane. The HIV-1 accessory protein Vpu counteracts BST-2 by downregulating it from the cell surface and displacing it
Envelope proteins of spleen necrosis virus form infectious human immunodeficiency virus type 1 pseudotype vector particles, but fail to incorporate upon substitution of the cytoplasmic domain with that of Gibbon ape leukemia virus.
Stitz, J; Wolfrum, N; Buchholz, CJ; Cichutek, K
The Journal of General Virology null
John D Lapek et al.
Molecular & cellular proteomics : MCP, 16(8), 1447-1461 (2017-06-14)
The mechanisms by which human immunodeficiency virus (HIV) circumvents and coopts cellular machinery to replicate and persist in cells are not fully understood. HIV accessory proteins play key roles in the HIV life cycle by altering host pathways that are
Anje Sporbert et al.
PloS one, 8(5), e64023-e64023 (2013-05-24)
This study describes a simple technique that improves a recently developed 3D sub-diffraction imaging method based on three-photon absorption of commercially available quantum dots. The method combines imaging of biological samples via tri-exciton generation in quantum dots with deconvolution and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service