Skip to Content
Merck
All Photos(1)

Key Documents

MAB8259

Sigma-Aldrich

Anti-Influenza B Antibody, nucleoprotein, clone B2

clone B2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
B2, monoclonal
application:
IF
species reactivity:
human
technique(s):
immunofluorescence: suitable
citations:
2

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

B2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

Application

This Anti-Influenza B Antibody, nucleoprotein, clone B2 is validated for use in IF for the detection of Influenza B.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Landgraf et al.
MethodsX, 8, 101442-101442 (2021-08-26)
We applied a one-step reverse transcriptase real-time PCR (rRT-PCR) analysis using TaqMan technique to evaluate the infectious titers of vaccine strains containing in trivalent live influenza vaccines (LAIVs). The cold-adapted reassortant influenza viruses A/H1N1 pdm09, A/H3N2, B/Yamagata and B/Victoria, included
Chung-Ming Yu et al.
Scientific reports, 10(1), 13318-13318 (2020-08-10)
Immunoassays based on sandwich immuno-complexes of capture and detection antibodies simultaneously binding to the target analytes have been powerful technologies in molecular analyses. Recent developments in single molecule detection technologies enable the detection limit of the sandwich immunoassays approaching femtomolar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service