Skip to Content
Merck
All Photos(1)

Key Documents

MAB805

Sigma-Aldrich

Anti-Adenovirus (Blend) Coating Antibody, clone 2/6, and 20/11

ascites fluid, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

2/6, monoclonal
20/11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1κ

shipped in

wet ice

General description

Adenoviruses are 65-80 nm, non-enveloped, regular icosahedron pathogens often associated with respiratory and gastrointestinal illness as well as conjunctivitis. Well over 40 types of adenoviruses have currently been recognized. In addition to their deleterious effects, adenoviruses have been used in vaccine production and in gene therapy. They also have the ability to trigger cell proliferation by interfering with host cellular anti-oncogenes. Since they are easy to prepare, multiple quickly and efficiently, and have the ability to infect and deliver their genome (including researcher-created constructs) to the nucleus of a variety of post-mitotic cells, adenoviruses are extremely valuable in research and medical applications ranging from virus/host cell interaction to the insertion of novel genes or regulatory mechanisms into eukaryotic cellular systems, to delivery of therapeutic anti-cancer genes.

Specificity

Reactive with all 41 serotypes of Adenovirus.

Immunogen

Adeno 3

Application

Anti-Adenovirus (Blend) Coating Antibody, clone 2/6 & 20/11 is an antibody against Adenovirus (Blend) Coating for use in ELISA, IF, IH, IH(P).
EIA: 1:1000

IFA: 1:200

Immunohistochemistry: 1:400 - 1:1000 for formalin fixed paraffin-embedded or frozen tissue.

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Adenovirus Control Slides, Catalogue Number 5009-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Cs Yeung et al.
Future oncology (London, England), 14(22), 2265-2277 (2018-03-24)
Idelalisib (IDELA) treatment is associated with diarrhea/colitis (incidence of ∼15% grade ≥3). We performed a retrospective analysis of gastrointestinal biopsies from 29 patients treated with IDELA across nine clinical trials. A central core laboratory performed histopathologic review, immunohistochemistry, and droplet digital
Highly augmented cytopathic effect of a fiber-mutant E1B-defective adenovirus for gene therapy of gliomas.
N Shinoura, Y Yoshida, R Tsunoda, M Ohashi, W Zhang, A Asai, T Kirino, H Hamada
Cancer Research null
Intravascular injections of a conditional replicative adenovirus (adl118) prevent metastatic disease in human breast carcinoma xenografts.
A Fabra, C Parada, A Vinyals, P Martin Duque, V Fernandez, R Sanchez-Prieto, S Ramon y Cajal
Gene Therapy null
Camden R Bair et al.
Journal of virology, 96(5), e0109021-e0109021 (2021-07-08)
Human adenovirus type 4 (HAdV-E4) is the only type (and serotype) classified at present within species Human mastadenovirus E that has been isolated from a human host. Recent phylogenetic analysis of whole-genome sequences of strains representing the spectrum of intratypic
Successful outcome of severe adenovirus hepatitis of the allograft following liver transplantation.
D Perez, L McCormack, H Petrowsky, W Jochum, B Mullhaupt, P-A Clavien
Transplant Infectious Disease : An Official Journal of the Transplantation Society null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service