Skip to Content
Merck
All Photos(1)

Documents

MAB5216

Sigma-Aldrich

Anti-NMDAR2A Antibody

Chemicon®, from mouse

Synonym(s):

NR2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

Immunogen

Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein.

Application

Anti-NMDAR2A Antibody detects level of NMDAR2A & has been published & validated for use in IH(P), IC, IH & WB.
Immunohistochemistry (light and EM) on 4% paraformaldehyde & 0.125% glutaraldehyde fixed tissue: 2.5-5 μg/mL.

Immunocytochemistry: 2.5-5.0μg/mL

Western blot: 1-5 μg/mL Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Linkage

Replaces: MAB5218

Physical form

Affinity purified immunoglobulin. Liquid in PBS. Contains no preservative.
Format: Purified

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruifa Mi et al.
Neuron, 44(2), 335-349 (2004-10-12)
Under standard conditions, cultured ventral spinal neurons cluster AMPA- but not NMDA-type glutamate receptors at excitatory synapses on their dendritic shafts in spite of abundant expression of the ubiquitous NMDA receptor subunit NR1. We demonstrate here that the NMDA receptor
Andre Fischer et al.
Neuron, 48(5), 825-838 (2005-12-13)
While deregulation of cyclin-dependent kinase 5 (Cdk5) has been implicated in neurodegenerative diseases, its precise role in synaptic plasticity and memory remains elusive. Proteolytic cleavage of p35, a regulatory subunit of Cdk5, by calpain results in the generation of the
Regulation of NMDA receptors by cyclin-dependent kinase-5.
Li, BS; Sun, MK; Zhang, L; Takahashi, S; Ma, W; Vinade, L; Kulkarni, AB; Brady, RO; Pant, HC
Proceedings of the National Academy of Sciences of the USA null
Qifa Li et al.
Frontiers in molecular neuroscience, 12, 26-26 (2019-03-12)
Chondroitin sulfate (CS) and dermatan sulfate (DS) proteoglycans (PGs) are major extracellular matrix (ECM) components of the central nervous system (CNS). A large body of evidence has shown that CSPGs/DSPGs play critical roles in neuronal growth, axon guidance, and plasticity
Céline Bidoret et al.
Frontiers in synaptic neuroscience, 7, 1-1 (2015-03-10)
N-methyl-D-aspartate receptors (NMDARs) in cerebellar molecular layer interneurons (MLIs) are expressed and activated in unusual ways: at parallel fibre (PF) synapses they are only recruited by repetitive stimuli, suggesting an extrasynaptic location, whereas their activation by climbing fibre is purely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service