Skip to Content
Merck
All Photos(1)

Key Documents

MAB4418

Sigma-Aldrich

Anti-Achaete Scute homolog 2 Antibody, clone 7E2

clone 7E2, from mouse

Synonym(s):

achaete-scute complex (Drosophila) homolog-like 2, achaete-scute complex homolog 2 (Drosophila), achaete-scute complex homolog-like 2, achaete-scute complex-like 2, achaete-scute complex-like 2 (Drosophila), mammalian achaete/scute homologue 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7E2, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASCL2(430)

General description

Achaete Scute homolog 2 (also known as mammalian aschaete-scute homolog 2 or Mash2) is a nuclear protein expressed specifically in the extravillous trophoblasts of developing placenta. Placental development involves control by the basic helix-loop-helix transcription factor Achaete Scute homolog 2, which may regulate HIF (hypoxia) in the formation of spongiotrophoblasts. Targeted mutagenesis of Achaete Scute homolog 2 yielded loss of function results in embryonic lethality at midgestation due to placental failure associated with a lack of spongiotrophoblasts and reduced labyrinthine trophoblast layers. In addition to healthy placental development, Achaete Scute proteins may also be involved in the determination of neuronal precursors in the central and peripheral nervous systems. Achaete Scute homolog 2 has also been implicated as a controller of intestinal stem cell fate and is essential for the maintenance of adult intestinal stem cells (Tanaka, M., et al., (1997) Dev Biol. 190(10):55-65).

Specificity

This antibody recognizes Achaete Scute homolog 2 near the C-terminus.

Immunogen

His-tagged recombinant protein corresponding to human Achaete Scute homolog 2 near the C-terminus.

Application

Anti-Achaete Scute homolog 2 Antibody, clone 7E2 detects level of Achaete Scute homolog 2 & has been published & validated for use in WB, IC, ChIP.
Immunocytochemmistry Analysis: A previous lot was used by an independent laboratory in IC (Van de Flier, L.G., et al., 2009)

Chromatin Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in ChIP (Hatzis, P., et al., 2008).

Quality

Evaluated by Western Blot in human placenta tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Achaete Scute homolog 2 in 10 µg of human placenta tissue lysate.

Target description

~20 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ye et al.
Cell cycle (Georgetown, Tex.), 17(8), 1014-1025 (2018-06-12)
The Wnt signaling pathway controls stem cell identity in the intestinal epithelium and cancer stem cells (CSCs). The transcription factor Ascl2 (Wnt target gene) is fate decider of intestinal cryptic stem cells and colon cancer stem cells. It is unclear
Yangyang Shang et al.
Oncotarget, 6(31), 30993-31006 (2015-08-27)
The role of Achaete scute-like 2 (Ascl2) in colorectal cancer (CRC) cell differentiation is unknown. LS174T, HT-29 and Caco-2 cells have high Ascl2 expression, while Lovo and SW480 cells have low Ascl2 expression. LS174T and HT-29 cells with Ascl2 knockdown
Genta Nagae et al.
Nature communications, 12(1), 5423-5423 (2021-09-21)
Hepatoblastoma (HB) is the most common pediatric liver malignancy; however, hereditary predisposition and acquired molecular aberrations related to HB clinicopathological diversity are not well understood. Here, we perform an integrative genomic profiling of 163 pediatric liver tumors (154 HBs and
Jarom Heijmans et al.
Cell reports, 3(4), 1128-1139 (2013-04-03)
Stem cells generate rapidly dividing transit-amplifying cells that have lost the capacity for self-renewal but cycle for a number of times until they exit the cell cycle and undergo terminal differentiation. We know very little of the type of signals
Kylie Hin-Man Mak et al.
Scientific reports, 11(1), 884-884 (2021-01-15)
Trophoblast stem cell (TSC) is crucial to the formation of placenta in mammals. Histone demethylase JMJD2 (also known as KDM4) family proteins have been previously shown to support self-renewal and differentiation of stem cells. However, their roles in the context

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service