Skip to Content
Merck
All Photos(1)

Key Documents

MAB3540

Sigma-Aldrich

Anti-Lamin A Antibody, CT, a.a. 598-611, clone 133A2

clone 133A2, Chemicon®, from mouse

Synonym(s):

70 kDa Lamin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

133A2, monoclonal

species reactivity

rat, mouse, canine, bovine, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG3

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Nuclear lamins form a network of filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished: A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2.

Specificity

Monoclonal 133A2 reacts against human lamin A but not lamin C. The epitope lies with the 98 amino acids of the C terminus that is unique to lamin A. Specifically deletion analysis has shown that amino acids 598-611 were essential for reactivity {Hozak, P et.al. (1995), J. Cell Sci 635-644}.

Immunogen

Epitope: C-terminus, a.a. 598-611
Synthetic peptide from the C-terminus of human lamin A. This sequence is not present in the the lamin C isoform.

Application

Immunoblotting: Lamin A detected as a 70kDa protein under reduced conditions.

Immunocytochemistry on fixed cells (methanol/acetone fixation)

Immunohistochemistry on frozen tissue sections.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Lamin A Antibody, C-terminus, a.a. 598-611, clone 133A2 is validated for use in FC, WB, IC, IH for the detection of Lamin A.

Physical form

Format: Purified
Liquid in buffer containing 0.1% sodium azide.

Storage and Stability

Maintain -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raju Padiya et al.
PloS one, 9(5), e94228-e94228 (2014-05-07)
Cardiovascular complication due to diabetes has remained a major cause of death. There is an urgent need to intervene the cardiac complications in diabetes by nutritional or pharmacological agents. Thus the present study was designed to find out the effectiveness
Micah J Drummond et al.
Journal of applied physiology (Bethesda, Md. : 1985), 111(1), 135-142 (2011-04-30)
Amino acid transporters and mammalian target of rapamycin complex 1 (mTORC1) signaling are important contributors to muscle protein anabolism. Aging is associated with reduced mTORC1 signaling following resistance exercise, but the role of amino acid transporters is unknown. Young (n
Petra Sehnalová et al.
Biology of the cell, 106(5), 151-165 (2014-03-13)
The optimal repair of DNA lesions is fundamental for physiological processes. We asked whether the recruitment of HP1β, 53BP1 and BMI1 proteins to ultraviolet (UVA)-induced DNA lesions requires functional A-type lamins. We found that UVA irradiation of nuclear lamina abolished
Jason P Chua et al.
Human molecular genetics, 23(5), 1376-1386 (2013-10-24)
Spinobulbar muscular atrophy (SBMA) is an inherited neuromuscular disorder caused by the expansion of a CAG repeat encoding a polyglutamine tract in exon 1 of the androgen receptor (AR) gene. SBMA demonstrates androgen-dependent toxicity due to unfolding and aggregation of
Laia Gómez-Baldó et al.
Cell cycle (Georgetown, Tex.), 9(6), 1143-1155 (2010-03-20)
Studies of the role of tuberous sclerosis complex (TSC) proteins (TSC1/TSC2) in pathology have focused mainly on their capacity to regulate translation and cell growth, but their relationship with alterations of cellular structures and the cell cycle is not yet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service