Skip to Content
Merck
All Photos(1)

Documents

MAB3418A5

Sigma-Aldrich

Anti-MAP2 Antibody, clone AP20, Alexa Fluor 555 Conjugate

clone AP20, from mouse, ALEXA FLUOR 555

Synonym(s):

Microtubule-associated protein 2, MAP-2, MAP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

AP20, monoclonal

species reactivity

rat, Xenopus, bovine, chicken, human, quail, mouse

technique(s)

immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

chicken ... Map2(424001)
frog ... Map2(780043)
human ... MAP2(4133)
mouse ... Map2(17756) , Map2(281294)
rat ... Map2(25595)

General description

The microtubule-associated protein 2 (MAP-2) is an abundant neuronal cytoskeletal protein that binds to tubulin, stabilizes microtubules (Herzog and Weber, 1978), and is an excellent marker for neurons. MAP-2 is essential for the development and maintenance of neuronal morphology (Matus, 1991). Three primary isoforms of MAP-2, the high molecular weight MAP2a and MAP2b, and the low molecular weight MAP2c (resulting from alternate splicing of the MAP2 gene), have been identified in neurons. (Chung et al., 1996). The low molecular weight isoform, MAP2c, is expressed in developing brain and is down-regulated during brain maturation, whereas the high molecular weight MAP2b is expressed in both developing and adult brain. The MAP2a appears only after brain maturation (Tucker, 1990). All these forms bind to microtubules through a domain near the carboxyl terminus that contains either three or four similar repeats of a 31-amino-acid motif (Lewis et al., 1988). MAP-2 together with MAP-4 and tau proteins belong to the family of thermostable proteins associated with microtubules.

Application

This Anti-MAP2 Antibody, clone AP20, Alexa Fluor 555 Conjugate is validated for use in ICC for the detection of MAP2.

Quality

Evaluated by Immunocytochemistry in rat E18 primary cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected MAP2 in rat E18 primary cortex cells.

Target description

~230 kDa observed.

Physical form

Purified mouse monoclonal IgG conjugated to Alexa Fluor 555 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Bosc et al.
Cell death & disease, 13(11), 959-959 (2022-11-16)
Caspase-2 (Casp2) is a promising therapeutic target in several human diseases, including nonalcoholic steatohepatitis (NASH) and Alzheimer's disease (AD). However, the design of an active-site-directed inhibitor selective to individual caspase family members is challenging because caspases have extremely similar active
Agnieszka Rybak-Wolf et al.
Nature microbiology, 8(7), 1252-1266 (2023-06-23)
Herpes simplex encephalitis is a life-threatening disease of the central nervous system caused by herpes simplex viruses (HSVs). Following standard of care with antiviral acyclovir treatment, most patients still experience various neurological sequelae. Here we characterize HSV-1 infection of human
Pauline Vaur et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(12), 5440-5452 (2017-08-27)
NAD+ depletion is a common phenomenon in neurodegenerative pathologies. Excitotoxicity occurs in multiple neurologic disorders and NAD+ was shown to prevent neuronal degeneration in this process through mechanisms that remained to be determined. The activity of nicotinamide riboside (NR) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service