Skip to Content
Merck
All Photos(2)

Key Documents

MAB341

Sigma-Aldrich

Anti-GABA A Receptor β 2,3 Chain Antibody, clone BD17

clone BD17, Chemicon®, from mouse

Synonym(s):

GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor, beta 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BD17, monoclonal

species reactivity

mouse, feline, monkey, bovine

species reactivity (predicted by homology)

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABRB2(2561)

General description

Gamma-aminobutyric acid type A (GABAA) receptors are members of the neurotransmitter ligand-gated ion channels that mediate neuronal inhibition on binding GABA. The effects of GABA on GABAA receptors are modulated by a range of therapeutically important drugs, including barbiturates, anaesthetics and benzodiazepines.

Specificity

Reacts with the β-chain of the GABAA receptor.

Immunogen

Epitope: Extracellular domain of the beta2 & beta3 subunits of the GABAA receptor
Purified GABA/benzodiazepine receptor from bovine cortex

Application

Detect GABA A Receptor β 2 using this Anti-GABA A Receptor β 2, 3 Chain Antibody, clone BD17 validated for use in IH, IP & WB.
Immunohistochemistry:
A 10-20 μg/mL concetration of a previous lot was used in IH.

Immunoprecipitation:
A previous lot of this antibody was used in IP.

Western blot:
5-20 µg/mL. Recognizes a 55 kDa band corresponding to the beta chain of the GABAA receptor.

Optimal working dilutions must be determined by end user.

Quality

Evaluated by Western Blot on mouse brain membrane lysates.

Western Blot:
1:500 dilution of this antibody detected GABA A Receptor β 2,3 Chain on 10 μg of mouse brain membrane lysates.

Target description

55 kDa

Linkage

Replaces: AB5952

Physical form

Format: Purified
Purified mouse monoclonal IgG1 liquid in buffer containing 10 mM potassium phosphate buffer pH 7.4 with 150 mM NaCl

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human ?1?3?2L gamma-aminobutyric acid type A receptors: High-level production and purification in a functional state.
Dostalova, Z; Zhou, X; Liu, A; Zhang, X; Zhang, Y; Desai, R; Forman, SA; Miller, KW
Protein Science null
Daniel C Adusei et al.
Neuropharmacology, 59(3), 167-171 (2010-05-18)
Fragile X syndrome is the most common heritable form of mental retardation. It is caused by silencing of the Fmr1 gene and the absence of the encoded protein. The purpose of this study was to examine global protein expression levels
Gamma-aminobutyric acid (GABA) is an autocrine excitatory transmitter in human pancreatic beta-cells.
Braun, M; Ramracheya, R; Bengtsson, M; Clark, A; Walker, JN; Johnson, PR; Rorsman, P
Diabetes null
Pengju Ma et al.
International journal of clinical and experimental medicine, 8(8), 14296-14302 (2015-11-10)
This study aimed to investigate GABA effects on blood pressure and blood dynamics of anesthetic rats by observing spontaneously hypertensive rats under both anesthesia and waking state. 72 male waking Wistar-Kyokos (WKY) rats and 72 male anesthetized spontaneously hypertensive (SHR)
Mirko Feldmann et al.
Cerebral cortex (New York, N.Y. : 1991), 29(2), 892-905 (2018-12-12)
Although by adulthood cortical structures and their capacity for processing sensory information have become established and stabilized, under conditions of cortical injury, or sensory deprivation, rapid reorganization occurs. Little is known as to the impact of this kind of adaptation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service