Skip to Content
Merck
All Photos(4)

Key Documents

MAB3408

Sigma-Aldrich

Anti-β-Tubulin III Antibody

CHEMICON®, mouse monoclonal, KMX-1

Synonym(s):

Tubulin beta-III, tubulin, beta 3, tubulin, beta 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KMX-1, monoclonal

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)

General description

Microtubules are constituent parts of the mitotic apparatus, cilia, flagella, and elements of the cytoskeleton. They consist principally of 2 soluble proteins, alpha- and beta-tubulin, each of about 55,000 kDa. Antibodies against beta Tubulin are useful as loading controls for Western Blotting. However it should be noted that levels of beta Tubulin may not be stable in certain cells. For example, expression of tubulin in adipose tissue is very low and therefore beta Tubulin should not be used as loading control for these tissues.

Specificity

This antibody reacts specifically with β-tubulin and shows a strong cross-reactivity to different species. This antibody allows staining of microtubuli using indirect immunofluorescence showing a low background only.

Immunogen

Tubulin purified from polycephalum myxamoebae

Application

Immunocytochemistry:
A 1-2 μg/mL concentration of a previous lot was used in immunocytochemistry.

Western blot:
0.05-0.5 μg/mL

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Tubulin Antibody, beta, clone KMX-1 is validated for use in IC, WB for the detection of Tubulin.

Quality

Routinely evaluated by Western Blot on A431 lysates.

Western blot:
1:500 dilution of this lot detected TUBULIN (BETA) on 10 μg of A431 lysates.

Target description

50 kDa

Linkage

Replaces: 04-1049

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2b liquid in buffer containing 0.02M phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8ºC in undiluted aliquots from date of receipt.

Analysis Note

Control
HeLa Cell lysate

A431 Cell lysate

MCF7 Cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard J Flannery et al.
Frontiers in synaptic neuroscience, 4, 6-6 (2012-12-12)
N-cadherin is a calcium-sensitive cell adhesion molecule commonly expressed at synaptic junctions and contributes to formation and maturation of synaptic contacts. This study used heterologous cell cultures of brainstem cholinergic neurons and transfected Chinese Hamster Ovary (CHO) cells to examine
Juan L Brusés
PloS one, 8(4), e62435-e62435 (2013-04-30)
Neuronal nicotinic acetylcholine receptors (nAChRs) are widely expressed throughout the central and peripheral nervous system and are localized at synaptic and extrasynaptic sites of the cell membrane. However, the mechanisms regulating the localization of nicotinic receptors in distinct domains of
Contactin-1 regulates myelination and nodal/paranodal domain organization in the central nervous system.
Colako?lu, G; Bergstrom-Tyrberg, U; Berglund, EO; Ranscht, B
Proceedings of the National Academy of Sciences of the USA null
Ari M Chow et al.
Cell stress & chaperones, 18(2), 155-160 (2012-08-07)
Alzheimer's disease, Parkinson's disease and amyotrophic lateral sclerosis (ALS) are 'protein misfolding disorders' of the mature nervous system that are characterized by the accumulation of protein aggregates and selective cell loss. Different brain regions are impacted, with Alzheimer's affecting cells
Ari M Chow et al.
Cell stress & chaperones, 19(6), 845-851 (2014-04-05)
Hsp70, Hsp32, and Hsp27 were induced by celastrol in rat cerebral cortical cultures at dosages that did not affect cell viability. Pronounced differences were observed in the cellular localization of these heat shock proteins in cell types of cerebral cortical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service