Skip to Content
Merck

MAB3402

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5

clone GA5, Chemicon®, from mouse

Synonym(s):

GFAP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
GA5, monoclonal
Application:
ICC, IHC (p), WB
Citations:
414
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GA5, monoclonal

species reactivity

chicken, pig, rat, bovine, mouse, rabbit, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

General description

50 kDa
Glial fibrillary acidic protein is a class-III intermediate filament. GFAP is the main constituent of intermediate filaments in astrocytes and serves as a cell specific marker that distinguishes differentiated astrocytes from other glial cells during the development of the central nervous system.

Immunogen

Purified Glial Fibrillary Acidic Protein (GFAP) from pig spinal cord (Debus, E., 1983).

Application

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 is an antibody against Glial Fibrillary Acidic Protein for use in IC, IH, IH(P) & WB with more than 55 product citations.
Immunocytochemistry:
5 μg/mL of a previous lot was used.

Immunohistochemistry:
5 µg/mL

Immunoblotting:
Recognizes a 51kDa protein in reducing westerns of total brain lysates.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Biochem/physiol Actions

Expected to cross-react with porcine, chicken, bovine, and rabbit.
The antibody reacts with GFAP from human, pig, chicken and rat. In tissue sections this antibody stains astrocytes and Bergman glia cells (Debus, E., 1983).

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 in buffer containing 0.02 M phosphate buffer, 0.25 M NaCl with 0.1% sodium azide, pH 7.6.

Preparation Note

Store the reconstituted antibody at 2-8°C for up to 6 months after date of receipt.
DO NOT FREEZE.

Analysis Note

Control
Mouse brain tissue, Astrocyte culture.
Routinely evaluated by Western Blot on Mouse brain lysate.

Western Blot Analysis:
1:1000 dilution of this lot detected Glial Fibrillary Acidic Protein on 10 μg of Mouse Brain lysate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library