Skip to Content
Merck
All Photos(1)

Key Documents

MAB2076Z

Sigma-Aldrich

Anti-Integrin beta 6 (ITGB6) Antibody

CHEMICON®, mouse monoclonal, CSbeta6

Synonym(s):

MAB2076

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin β6 Antibody, clone CSβ6, azide free, clone CSbeta6, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CSbeta6, monoclonal

species reactivity

human, monkey, rat, rabbit

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB6(3694)
mouse ... Itgb6(16420)
rat ... Itgb6(311061)
rhesus monkey ... Itgb6(700454)

Related Categories

Specificity

Monoclonal antibody MAB2076Z, raised by immunizing beta6 knockout mice with recombinant secreted human alphavbeta6, is useful for flow cytometry and immunoprecipitation in all species tested, including mouse. It also works well for immunohistochemistry of acetone-fixed tissues in several species, including rat, rabbit, monkey, and human. The antibody does not work well for immunohistochemistry of mouse tissue. This antibody does not recognize the aV subunit and does not react with alphaV integrins other than alphavbeta6. Because the antibody is not effective in western blot, it is unknown whether the epitope recognized by the antibody localizes to the alphavbeta6 heterodimer or to the beta6 subunit only.

Immunogen

beta 6 knockout mice have been immunized with recombinant secreted human alpaVbeta6.

Application

Anti-Integrin β6 Antibody, clone CSβ6, azide free detects level of Integrin β6 & has been published & validated for use in FC, IP, IC, IH.
Immunohistochemistry (acetone fixation): Effective for Rabbit, rat, monkey, and human. Not effective for mouse tissue.

Immunocytochemistry: Effective for F6, HT-29, H441 cell lines (others not tested).

Flow cytometry: Effective for all species tested, including rabbit, rat, monkey, human, and mouse.

Immunoprecipitation

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Sterile-filtered liquid at 1 mg/mL in PBS, containing no preservatives.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A continuous bovine kidney cell line constitutively expressing bovine ?v?6 integrin has increased susceptibility to foot-and-mouth disease virus.
LaRocco, M; Krug, PW; Kramer, E; Ahmed, Z; Pacheco, JM; Duque, H; Baxt, B; Rodriguez, LL
Journal of Clinical Microbiology null
A liposomal drug platform overrides peptide ligand targeting to a cancer biomarker, irrespective of ligand affinity or density.
Gray, BP; McGuire, MJ; Brown, KC
Testing null
Novel ITGB6 variants cause hypoplastic-hypomineralized amelogenesis imperfecta and taurodontism: characterization of tooth phenotype and review of literature.
Sriwattanapong, et al.
BDJ open, 9, 15-15 (2023)
Yahui Jiang et al.
Frontiers in oncology, 11, 690618-690618 (2021-10-09)
Implantation metastasis is the main means of dissemination in ovarian cancer. Our previous studies showed that SET and MYND domain-containing protein 3 (SMYD3) expression was higher in ovarian cancer spheroids than in monolayers. SMYD3 enhancement of spheroid invasion and adhesion
Dana L Nettles et al.
Journal of anatomy, 204(6), 515-520 (2004-06-17)
In this study, we investigated the profile of integrin expression in human and porcine intervertebral disc tissue. Differences in extracellular matrix composition between anulus fibrosus (AF) and nucleus pulposus (NP) regions of the disc, as well as differences in cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service