Skip to Content
Merck
All Photos(1)

Documents

MAB2060

Sigma-Aldrich

Anti-Integrin β4 Antibody, clone ASC-9

clone ASC-9, Chemicon®, from mouse

Synonym(s):

CD104

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ASC-9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB4(3691)

Specificity

Human beta4 integrin (CD104)

Immunogen

Human squamous cell carcinoma cells (SCC9)

Application

Inhibition of cell attachment: inhibits SCC9 cell adhesion to human and

EHS laminin, but not to laminin GD-2 peptide, type IV collagen or fibronectin. Does not inhibit SKOV-3 cell adhesion to laminin, fibronectin or type IV collagen.

Immunohistochemistry: on acetone-fixed frozen human epithelial tissues including tongue, foreskin and ovary. Not for use on formalin-fixed tissue.

Flow cytometry: stains human squamous cell carcinoma (SCC9), ovarian carcinoma (SKOV-3) and umbilical vein endothelium (HUVEC). Stains RKO or K562 cells transfected for CD104 expression. Does not stain monocytes, neutrophils, lymphocytes, T cells, B cells or platelets.

Immunoprecipitation: precipitates PAGE bands of 66, 76, 120, 135 and 200kDa (reduced) or 66, 110, 125, 130 and 190kDa (non-reduced) from surface-biotinylated SCC9 cells.

Working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin β4 Antibody, clone ASC-9 is validated for use in FC, IP, IH, FUNC for the detection of Integrin β4.

Physical form

Format: Purified
Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan M Fennewald et al.
The Journal of biological chemistry, 287(25), 21058-21066 (2012-05-02)
Lymphatic metastasis of cancer cells involves movement from the primary tumor site to the lymph node, where the cells must be able to productively lodge and grow. It is there that tumor cells encounter cellular and non-cellular constituent elements that
Yukinori Endo et al.
Cell adhesion & migration, 12(2), 93-100 (2016-10-08)
Anosmin is an extracellular matrix protein, and genetic defects in anosmin result in human Kallmann syndrome. It functions in neural crest formation, cell adhesion, and neuronal migration. Anosmin consists of multiple domains, and it has been reported to bind heparan
J Nevo et al.
Oncogene, 29(49), 6452-6463 (2010-08-31)
The majority of mortality associated with cancer is due to formation of metastases from the primary tumor. Adhesion mediated by different integrin heterodimers has an important role during cell migration and invasion. Protein interactions with the β1-integrin cytoplasmic tail are
Role of integrins in the assembly and function of hensin in intercalated cells.
Vijayakumar, S; Erdjument-Bromage, H; Tempst, P; Al-Awqati, Q
Journal of the American Society of Nephrology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service