Skip to Content
Merck
All Photos(2)

Key Documents

MAB1997

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

Anti-Integrin β1 Antibody, clone MB1.2

CHEMICON®, rat monoclonal, MB1.2

Synonym(s):

CD29 antigen, Fibronectin receptor subunit beta, Integrin VLA-4 subunit beta, fibronectin receptor beta subunit, integrin VLA-4 beta subunit, integrin beta 1, integrin beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Integrin β1 Antibody, clone MB1.2, clone MB1.2, Chemicon®, from rat

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MB1.2, monoclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Integrin beta-1 (UniProt P05556; also known as CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12, VLA-BETA, VLAB) gene (Gene ID 3688) in human. Integrin beta-1 is a 130 kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.

Specificity

Reacts with β1 subunit of VLA (β1) integrins

Immunogen

Mouse Integrin beta-1, as per (Von Ballestrem, C.G., et al. (1996). Hybridoma. 15(2):125-132). Mouse Integrin beta-1, as per (Von Ballestrem, C.G., et al. (1996). Hybridoma. 15(2):125-132).

Application

Anti-Integrin β1 Antibody, clone MB1.2 is an antibody against Integrin β1 for use in FC, IH, IP & WB.
Immunoprecipitation:
A previous lot of this anitbody was used to immunoprecipitate VLA (β1) integrins from lysate of 106 cell equivalent: 2 μg.

Flow cytometry:
A 10 μg/mL concentration of a previous lot was used in FC.

Immunohistochemistry in frozen tissue sections:
20 μg/mL from a previous lot was used. As a result of the wide tissue distribution of VLA (β1), binding of MAB1997 to diverse cell types is commonly observed.
Does not block VLA integrin mediated cell adhesion to matrix proteins. Activation of VLA (β1) integrin signaling properties by MAB1997 has not been characterized.

Western blot:
10 µg/mL. Effective for samples that have been SDS-denatured and heated. Not effective for reduced samples.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on C2C12 lysate.

Western Blot Analysis:
1:1000 dilution of this lot detected integrin β1 on 10 μg of C2C12 lysate.

Target description

~88 kDa (non-glycosylated) and ~130 kDa ( glycosylated).

Linkage

Replaces: 04-1109

Physical form

Format: Purified
Purified rat monoclonal IgG2κ liquid in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl with 0.1% NaN3

Analysis Note

Control
Human OVCAR5 cell line whole cell lysate

C2C12 lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dystroglycan does not contribute significantly to kidney development or function, in health or after injury.
Jarad, G; Pippin, JW; Shankland, SJ; Kreidberg, JA; Miner, JH
American Journal of Physiology: Renal Physiology null
Won-Jing Wang et al.
The Journal of cell biology, 159(1), 169-179 (2002-10-09)
Death-associated protein kinase (DAP-kinase) is a calcium/calmodulin-dependent serine/threonine kinase, and participates in various apoptosis systems. However, its apoptosis-promoting mechanism is poorly understood. Here, we demonstrate that DAP-kinase suppresses integrin-mediated cell adhesion and signal transduction, whereas dominant-negative interference of this kinase
Reversion of the Jun-induced oncogenic phenotype by enhanced synthesis of sialosyllactosylceramide (GM3 ganglioside).
Miura, Y; Kainuma, M; Jiang, H; Velasco, H; Vogt, PK; Hakomori, S
Proceedings of the National Academy of Sciences of the USA null
?Np63 knockout mice reveal its indispensable role as a master regulator of epithelial development and differentiation.
Romano, RA; Smalley, K; Magraw, C; Serna, VA; Kurita, T; Raghavan, S; Sinha, S
Development null
Maretoshi Hirai et al.
Genesis (New York, N.Y. : 2000), 54(5), 286-296 (2016-03-19)
Centrosomes are major microtubule organizing centers (MTOCs) that play an important role in chromosome segregation during cell division. Centrosomes provide a stable anchor for microtubules, constituting the centers of the spindle poles in mitotic cells, and determining the orientation of

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    幅広い条件で高感度にマウスIntegrin β1を染色可能。
    添付写真:マウス終脳(E10.5日)パラフィン切片
    ・固定:4%PFA/PBS(-)、4℃、O/N
    ・抗原賦活化:DAKO S1700、105℃、15分(121℃では抗原性が失われた)
     (PLP固定や10%TCA固定の凍結切片では、界面活性剤処理のみでOK)
    ・抗体反応:MB1.2 mAb @2 μg/ml(1 μg/mlでも十分だろう)、4℃、O/N

    Translate with Google

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service