Skip to Content
Merck
All Photos(1)

Documents

MAB1924

Sigma-Aldrich

Anti-Laminin α5 Antibody, clone 4C7

ascites fluid, clone 4C7, Chemicon®

Synonym(s):

Anti-AA408760, Anti-AA408762, Anti-AI853660, Anti-laminin-511, Anti-mKIAA0533

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4C7, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LAMA2(3908)

Specificity

Reacts with the globular domain of the alpha5 chain of intact laminin and blocks the neurite stimulating activity of laminin. This antibody was originally thought to recognize laminin alpha 1 or A, but further characterization has confirmed its specificity for the laminin alpha 5 chain [see Tiger, C.F. (1997) J. Biol. Chem. 272: 28590].

Application

Anti-Laminin α5 Antibody, clone 4C7 is an antibody against Laminin α5 for use in ELISA, IH & IP.
ELISA (50% maximal binding to human laminin): >1:11,000.

Immunofluorescence - tissue staining pattern is most consistent with that observed for laminin alpha5 in the mouse(Miner et al., 1997)

Affinity chromatography

Immunoprecipitation

Does not work for Western blotting

Optimal working dilutions must be determined by end user.

Physical form

UnPurified mouse ascites containing no preservatives.

Analysis Note

Control
Developing striated muscle tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamie L Maciaszek et al.
Biophysical journal, 102(5), 1137-1143 (2012-03-13)
Collapse and sudden death in physical training are the most serious complications of sickle cell trait (SCT). There is evidence that erythrocytes in SCT patients aggregate during strenuous exercise, likely because of adhesive interactions with the extracellular matrix (ECM) and
Giuseppe Mazza et al.
Scientific reports, 7(1), 5534-5534 (2017-07-19)
The development of human liver scaffolds retaining their 3-dimensional structure and extra-cellular matrix (ECM) composition is essential for the advancement of liver tissue engineering. We report the design and validation of a new methodology for the rapid and accurate production
Elena Daniele et al.
BMJ open ophthalmology, 7(1) (2022-09-27)
Recent clinical studies have shown that the transplantation of functional retinal pigment epithelium (RPE) cells can prevent the onset of RPE degeneration in age-related macular degeneration. This study aimed to investigate the potential of human amniotic membrane (hAM) as a
Mariko Shimizu et al.
Laboratory investigation; a journal of technical methods and pathology, 89(2), 178-195 (2008-12-17)
Nephrin is an essential structural component of the glomerular slit diaphragm (SD), a highly organized intercellular junction that constitutes the ultrafiltration barrier of the kidney. Recent studies have identified two additional nephrin-interacting SD proteins (NEPH1 and NEPH2), suggesting that the
Yuji Sekine et al.
Journal of the American Society of Nephrology : JASN, 20(7), 1586-1596 (2009-05-16)
LAT3 is a Na+-independent neutral l-amino acid transporter recently isolated from a human hepatocellular carcinoma cell line. Although liver, skeletal muscle, and pancreas are known to express LAT3, the tissue distribution and physiologic function of this transporter are not completely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service