Skip to Content
Merck
All Photos(1)

Key Documents

MAB1430

Sigma-Aldrich

Anti-Collagen Type IV Antibody, clone 24.12.8 (PHM-12)

clone 24.12.8 (PHM-12), Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

24.12.8 (PHM-12), monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

BIOCHEMISTRY:

p I = 5.5

Specificity

MAB1430 provides a marker for an antigen known to be important in glomerular cell-matrix interactions, and should prove useful in research into the mechanisms involved in renal pathology. This antibody reacts with glomerular and tubular basement membranes in kidney as well as basal lamina of capillaries. It also labels mesangial cells and matrix within the glomerulus as well as basement membrane structures in all organs examined (Gusterson et al., 1984).

Immunogen

Human glomeruli

Application

Detect Collagen Type IV using this Anti-Collagen Type IV Antibody, clone 24.12.8 (PHM-12) validated for use in IH, IH(P).
The antibody is suitable for use on both frozen and protease digested paraffin processed tissue.

SUGGESTED USAGE

Indirect immunoperoxidase staining - the final dilution will depend on the assay conditions and detection system employed. However, a dilution of at least 1:40 will be applicable to most commonly used systems.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin presented as a liquid in phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide.

Storage and Stability

Store at 2 to 8°C, for up to 6 months. For prolonged periods, store below -20°C in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphofunctional studies of the glomerular wall in mice lacking entactin-1.
Simon-Philippe Lebel, Yong Chen, Diane Gingras, Albert E Chung, Moise Bendayan
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society null
Invading squamous cell carcinoma can retain a basal lamina. An immunohistochemical study using a monoclonal antibody to type IV collagen.
Gusterson, B A, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 51, 82-87 (1984)
Production of monoclonal antibodies to fibronectin, type IV collagen and other antigens of the human glomerulus.
Hancock, W W, et al.
Pathology, 16, 197-206 (1984)
S E Aggrey et al.
Poultry science, 73(12), 1822-1828 (1994-12-01)
Records of 1,530 Japanese quail were used to estimate heritabilities and genetic correlations based on a derivative-free restricted maximum likelihood (REML) method with an animal model and ANOVA. The animal model included fixed effects of hatch and sex, random effects
Inflammation, immune reactivity, and angiogenesis in a severe combined immunodeficiency model of rheumatoid arthritis.
Davis, LS; Sackler, M; Brezinschek, RI; Lightfoot, E; Bailey, JL; Oppenheimer-Marks, N; Lipsky, PE
The American Journal of Pathology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service