Skip to Content
Merck
All Photos(2)

Key Documents

MAB131873

Sigma-Aldrich

Anti-mCherry Antibody, clone 1C51

clone 1C51, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C51, monoclonal

species reactivity (predicted by homology)

all

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

mCherry, red fluorescent protein, is derived from DsRed, a red fluorescent protein from disc corals of Discosoma species. It is monomeric form that has been engineered with multiple cycles of mutation, directed modification and evolutionary selection. It exhibits excitation maximum at 587 nm and an emission maximum at 610 nm with a fluorescent quantum yield of 0.22. mCherry displays high photostability and resistant to photobleaching. It also displays improved brightness and extremely rapid maturation rate. Anti-mCherry, clone1C51 is an affinity purified antibody that recognizes mCherry in HEK293 cells transfected with pFin-EF1-mCherry. (Ref.: Schaner, NC et al. (2004). Nat. Biotechnol. 22(12):1567-72).

Specificity

Clone 1C51 detects mCherry protein in multiple species.

Immunogen

His-tagged full length recombinant mCherry.

Application

Anti-mCherry Antibody, clone IC51, Cat. No. MAB131873 detects mCherry protein and is tested for use in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A representative lot detected mCherry in Western Blotting applications (Krajcovic, M., et. al. (2013). Mol Biol Cell. 24(23):3736-45).

Western Blotting Analysis: A representative lot detected mCherry in HEK293 cells transfected with pFin-EF1-mCherry vs HEK293 untransfected cells (Courtesy of Dr Gerry Shaw).

Immunocytochemistry Analysis: A representative lot detected mCherry in HEK293 cells transfected with pFin-EF1-mCherry (Courtesy of Dr Gerry Shaw).

Quality

Evaluated by Western Blotting in HEK293 cells transfected with pFin-EF1-mCherry.

Western Blotting Analysis: 1 µg/mL of this antibody detected mCherry in 10 µg of lysate from HEK293 cells transfected with pFin-EF1-mCherry.


Target description

~32 kDa Observed; 26.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG2a in buffer containing PBS with 0.03% sodium azide with 50% glycerol.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brittany M White-Mathieu et al.
Current protocols, 4(5), e1051-e1051 (2024-05-23)
Fluorescent imaging of cellular membranes is challenged by the size of lipid bilayers, which are smaller than the diffraction limit of light. Recently, expansion microscopy (ExM) has emerged as an approachable super-resolution method that requires only widely accessible confocal microscopes.
Nicole L Nuckolls et al.
PLoS genetics, 18(12), e1009847-e1009847 (2022-12-09)
Meiotic drivers bias gametogenesis to ensure their transmission into more than half the offspring of a heterozygote. In Schizosaccharomyces pombe, wtf meiotic drivers destroy the meiotic products (spores) that do not inherit the driver from a heterozygote, thereby reducing fertility.
Hongyan Sun et al.
iScience, 27(6), 110113-110113 (2024-07-02)
The gut epithelium is subject to constant renewal, a process reliant upon intestinal stem cell (ISC) proliferation that is driven by Wnt/β-catenin signaling. Despite the importance of Wnt signaling within ISCs, the relevance of Wnt signaling within other gut cell
N Quan et al.
Frontiers in genetics, 14, 1198203-1198203 (2023-09-25)
Protein misfolding is a common intracellular occurrence. Most mutations to coding sequences increase the propensity of the encoded protein to misfold. These misfolded molecules can have devastating effects on cells. Despite the importance of protein misfolding in human disease and
Matthew B Pomrenze et al.
Neuron, 110(24), 4125-4143 (2022-10-07)
Social isolation during opioid withdrawal is a major contributor to the current opioid addiction crisis. We find that sociability deficits during protracted opioid withdrawal in mice require activation of kappa opioid receptors (KORs) in the nucleus accumbens (NAc) medial shell.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service