Skip to Content
Merck
All Photos(1)

Key Documents

MAB1243

Sigma-Aldrich

Anti-Cortisol Antibody, 3-CMO, clone F4P1A3

clone F4P1A3, Chemicon®, from mouse

Synonym(s):

Anti-Cortisol antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F4P1A3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG2b

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Cortisol. 1 x 109 l/mol.

CrossReactivity:

Cortisone 100%

Prednisolone 45%

11-Deoxycortisol 55%

Immunogen

Cortisol-3-CMO-BSA
Epitope: 3-CMO

Application

Anti-Cortisol Antibody, 3-CMO, clone F4P1A3 is an antibody against Cortisol for use in ELISA.
Immunoassay.

Dilute with buffer pH 7.5 to 8.0 to desired working volumes.

Optimal working dilutions must be determined by end user.
Research Category
Metabolism
Research Sub Category
Metabolic Hormones & Receptors

Linkage

Replaces: CBL72

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS, pH 7.4 with 50% glycerol and 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to twelve months.Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service