Skip to Content
Merck

MA400

mA400 Basic Power Supply

Synonym(s):

Basic power supply

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105303
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 ea

General description

Basic power supply for protein and DNA electrophoresis

MA400-USmA400 Basic Power Supply, US plug
MA400-EUmA400 Basic Power Supply, Euro plug
MA400-UKmA400 Basic Power Supply, UK plug
MA400-NImA400 Basic Power Supply, Japan plug
MA400-ZHmA400 Basic Power Supply, China plug

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

  • For standard protein and DNA electrophoresis
  • Tank electroblotting up to 2 gels
  • Constant voltage or constant current
  • 2-step programs
  • Compact design

  • Gel Electrophoresis
  • Immunoblotting (Western)
  • Western Blotting





Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library