Skip to Content
Merck

JU-1L-BK

Blood typing Intermediates For Further Manufacturing Use (FFMU) - Anti Human Globulin

Anti-C3d from the clone BRIC-8, Murine IgM, Intermediate for further manufacturing use

Synonym(s):

Anti-C3d (clone BRIC-8), FFMU

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
BRIC-8, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-


biological source

mouse

Quality Segment

antibody form

unpurified

clone

BRIC-8, monoclonal

form

liquid

packaging

pack of 1 L

concentration

2-10 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline. For the formulation of Anti-Human Globulin blood typing reagents or devices. Detection of complement activation can aid in the diagnosis of autoimmune haemolytic anaemia, or the detection of some blood group allo-antibodies, particularly within the Kidd System. Contains 0.1% (w/v) Sodium Azide; Shelf Life 24 months from date of filtration. Licensed by the US FDA under a shared manufacturing arrangement.

Application

Antibody intermediate for further manufacturing use.
Production of an in vitro blood typing product or device.

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%<nbsp/>(w/v) sodium azide.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library