Skip to Content
Merck
All Photos(2)

Key Documents

HCVD3MAG-67K

Millipore

MILLIPLEX® Human Cardiovascular Disease (Acute Phase) Magnetic Bead Panel 3 - Cardiovascular Disease Multiplex Assay

The analytes available for this multiplex kit are: Adipsin, AGP, α2-Macroglobulin, CRP, Fetuin A, Fibrinogen, L-Selectin, Serum Amyloid P, Haptoglobin, & Platelet Factor-4.

Synonym(s):

human CVD APP immunoassay panel, luminex human cardiovascular disease acute phase proteins multiplex assay, millipore human CVD acute phase proteins multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 102%
(Haptoglobin)

accuracy: 109%
(AGP)

accuracy: 113%
(Platelet Factor-4)

accuracy: 117%
(Adipsin)

standard curve range: 0.004-15 ng/mL
(L-Selectin & SAP)

standard curve range: 0.012-50 ng/mL
(CRP)

standard curve range: 0.037-150 ng/mL
(PF4)

standard curve range: 0.061-250 ng/mL
(HPTGN)

standard curve range: 0.098-400 ng/mL
(AGP & Adipsin)

standard curve range: 0.244-1,000 ng/mL
(Fetuin A & vWF)

standard curve range: 0.488-2,000 ng/mL
(A2M)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cardiovascular disease, particularly atherosclerotic vascular disease, is a leading cause of global mortality, accounting for 30% of all global deaths (WHO, 2010). Inflammatory mechanisms play a vital role in the initiation, maintenance, and progression of vascular disease with a strong correlation between inflammatory markers and prognosis in acute and chronic coronary artery disease. Numerous studies have demonstrated an association of obesity and diabetes with cardiovascular risk factors.

MILLIPLEX® Human Cardiovascular Disease (CVD) Panel 3 (Acute Phase) is a 10-plex kit to be used for the simultaneous quantification of any or all of the following analytes in human serum, plasma or tissue/cell lysate and culture supernatant samples: Adipsin, α-1-Acid-Glycoprotein (AGP), α-2-Macroglobulin (A2M), C Reactive Protein (CRP), Fetuin A, Fibrinogen, L-Selectin, Serum Amyloid P (SAP), Haptoglobin, and Platelet Factor-4 (PF4). (Please note: Fibrinogen is not detectable in serum.) This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cardiovascular

Application

  • Analytes: α-1 Acid Glycoprotein (AGP), α-2-Macroglobulin (A2M), Adipsin (Factor D), C Reactive Protein (CRP), Fetuin A, Fibrinogen, Haptoglobin, sL-Selectin, Platelet Factor-4 (PF4/CXCL4), Serum Amyloid P (SAP)
  • Recommended Sample Type: Human serum, plasma, cell/tissue culture supernatants and lysates
  • Recommended Sample Dilution: 25 μL per well of 1:40,000 diluted serum or plasma; tissue/cell culture samples may require dilution in appropriate control medium
  • NOTE: Fibrinogen is not detectable in serum
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C or 2 hours at room temperature (20-25 °C)
  • Research Category: Cardiovascular Disease
  • Research Subcategory: Metabolic Disorders

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: See kit protocol for individual analyte sensitivity.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coagulation factors, fibrinogen and plasminogen activator inhibitor-1, are differentially regulated by yellow fever virus infection of hepatocytes.
Woodson, SE; Freiberg, AN; Holbrook, MR
Virus Research null
Anna C Belkina et al.
Frontiers in immunology, 9, 2783-2783 (2018-12-21)
Even with effective viral control, HIV-infected individuals are at a higher risk for morbidities associated with older age than the general population, and these serious non-AIDS events (SNAEs) track with plasma inflammatory and coagulation markers. The cell subsets driving inflammation
Yaochen Zhang et al.
PloS one, 12(7), e0182141-e0182141 (2017-07-28)
Fibrinogen, 1 of 13 coagulation factors responsible for normal blood clotting, is synthesized by hepatocytes. Detailed roles of the orphan nuclear receptors regulating fibrinogen gene expression have not yet been fully elucidated. Here, we identified estrogen-related receptor gamma (ERRγ) as
V B Silva et al.
Scientific reports, 9(1), 2386-2386 (2019-02-23)
Dialysate calcium concentration (d[Ca]) might have a cardiovascular impact in patients on haemodialysis (HD) since a higher d[Ca] determines better hemodynamic tolerability. We have assessed the influence of d[Ca] on global longitudinal strain (GLS) by two-dimensional echocardiography using speckle-tracking imaging
Daniel O'Connor et al.
Molecular systems biology, 16(11), e9888-e9888 (2020-11-20)
Neisseria meningitidis is a major cause of meningitis and septicaemia. A MenB vaccine (4CMenB) was licensed by the European Medicines Agency in January 2013. Here we describe the blood transcriptome and proteome following infant immunisations with or without concomitant 4CMenB

Related Content

Learn about biomarkers of cardiovascular disease (CVD) and discover how multiplex CVD panels, such as MILLIPLEX® multiplex assays, are being used to increase the efficiency of biomarker discovery for CVD research.

Learn about biomarkers of cardiovascular disease (CVD) and discover how multiplex CVD panels, such as MILLIPLEX® multiplex assays, are being used to increase the efficiency of biomarker discovery for CVD research.

Learn about biomarkers of cardiovascular disease (CVD) and discover how multiplex CVD panels, such as MILLIPLEX® multiplex assays, are being used to increase the efficiency of biomarker discovery for CVD research.

Learn about biomarkers of cardiovascular disease (CVD) and discover how multiplex CVD panels, such as MILLIPLEX® multiplex assays, are being used to increase the efficiency of biomarker discovery for CVD research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service