Skip to Content
Merck

Skip To

GPWP04700

PES Membrane Filter, 0.22 μm Pore Size

greener alternative

Greener Alternative

Millipore Express®, filter diam. 47 mm, hydrophilic

Synonym(s):

Hydrophilic polyethersulfone membrane filter discs, Millipore Express PLUS Membrane Filter, PES membrane filter discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.22 μm pore size
Material:
plain filter, polyethersulfone membrane, white filter
Feature:
hydrophilic
Diam.:
47 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

plain filter, polyethersulfone membrane, white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore Express®, Millipore

sustainability

Greener Alternative Product

parameter

15 mL/min water flow rate at 690 mbar (10 psi)

diam.

47 mm

filter diam.

47 mm

thickness

160-185 μm

impurities

≤0.5 EU/mL (LAL test, Aqueous extraction)

pore size

0.22 μm pore size

bubble point

≥1.38 bar, air with 100% IPA at 23 °C

greener alternative category

shipped in

ambient

General description

Filter Code: GPWP
Filter Type: Screen filter
The Millipore Express PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features & Benefit:
•Ultra fast filtration and high throughput shorten process time
•Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications

Applications:
Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.
Need a custom membrane filter? Click here to request a custom membrane filter or unique and large package sizes that may not be available via our website.

Application

  • Sterile filtration
  • Buffer filtration
  • Tissue culture media filtration
  • Sample filtration for PFAS analytical testing (LC-MS)

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MILLIPORE EXPRESS is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPWP04700HPWP09050GPWP01300
description

Millipore Express®, filter diam. 47 mm, hydrophilic

description

Millipore Express®, filter diam. 47 mm, hydrophilic

description

Millipore Express®, filter diam. 90 mm, hydrophilic

description

Millipore Express®, filter diam. 13 mm, hydrophilic

pore size

0.22 μm pore size

pore size

0.45 μm pore size

pore size

0.45 μm pore size

pore size

0.22 μm pore size

parameter

15 mL/min water flow rate at 690 mbar (10 psi)

parameter

23 mL/min water flow rate at 690 mbar (10 psi)

parameter

23 mL/min water flow rate at 690 mbar (10 psi)

parameter

15 mL/min water flow rate at 690 mbar (10 psi)

diam.

47 mm

diam.

47 mm

diam.

90 mm

diam.

13 mm

manufacturer/tradename

Millipore Express®

manufacturer/tradename

Millipore Express®

manufacturer/tradename

Millipore Express®

manufacturer/tradename

Millipore Express®

material

plain filter, white filter, polyethersulfone membrane

material

plain filter, polyethersulfone membrane, white filter

material

plain filter, polyethersulfone membrane, white filter

material

plain filter, polyethersulfone membrane, white filter

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters