GG-10X10ML-B
BIOSCOT™ IVD Reagent ABO & RHD (Rh) Specificities
BIOSCOT™ Anti-D from the clone RUM-1, Human IgM, Blood Grouping Reagent
Synonym(s):
Anti-D (clone RUM-1) Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
NACRES:
NA.41
Recommended Products
biological source
human
Quality Level
clone
RUM-1, monoclonal
packaging
pkg of 10 × 10 mL
manufacturer/tradename
BIOSCOT™
storage condition
do not freeze
concentration
25-40 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.6-7.0
isotype
IgM
application(s)
clinical testing
shipped in
ambient
storage temp.
2-8°C
General description
Appropriate for patient and neonatal testing - does not detect DVI red cells
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
Product formulation examined according to Annex IV of the EU IVD directive to support CE registration.
For In Vitro use only .
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
Product formulation examined according to Annex IV of the EU IVD directive to support CE registration.
For In Vitro use only .
Application
For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service