Skip to Content

GF302

BMP-4 Protein, Human Recombinant Animal Free

Bone morphogenetic protein 4 (BMP4) is a TGFβ superfamily lig& that is widely expressed from early embryogenesis through adulthood. The Recombinant animal free human BMP-4 protein is manufactured using all non-animal reagents.

Synonym(s):

BMP4, Bone morphogentic protein-4, DVR-4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Assay:
>98% (SDS-PAGE and HPLC)
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Bone morphogentic protein 4 (BMP4) is a TGFβ superfamily ligand that is widely expressed from early embryogenesis through adulthood. It plays an important role in mesenchyme formation, epidermal determination, suppression of neural induction, the development of multiple organs, and tissue repair. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva.
Animal free human recombinant BMP4 is a fully active homodimeric protein consisting of two 107 amino acid subunits which correspond to amino acids 303-408 of the full length BMP-4 precursor.
Product Source: E. coli

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Biochem/physiol Actions

Cross Reactivty
Mouse

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human BMP-4 is manufactured using all non-animal reagents

Preparation Note

Store at -20°C for up to 6 months from date of receipt.

Analysis Note

Bioactivity assay: Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 5-10 ng/ml.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library