Skip to Content
Merck
All Photos(1)

Key Documents

FP2

Millipore

mPAGE® Freezer Pack

for use with mPAGE® Mini Wet Transfer System

Synonym(s):

ice pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105305
NACRES:
NC.01

Pricing and availability is not currently available.

packaging

pkg of 1 pack

manufacturer/tradename

Millipore

technique(s)

immunoblotting: suitable
western blot: suitable

compatibility

for use with mPAGE® Mini Wet Transfer System

General description

mPAGE® Freezer Pack for use with mPAGE® Mini Wet Transfer System (MWTS).

The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service