Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB214P

Sigma-Aldrich

Milli-Mark® Anti-CD117 Antibody, c-kit-PE, clone 104D2

clone 104D2, Milli-Mark®, from mouse

Synonym(s):

Mast/stem cell growth factor receptor, SCFR, Tyrosine-protein kinase Kit, Proto-oncogene c-Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

104D2, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)
mouse ... Kit(16590)

Related Categories

General description

CD117 is a 145 kDa transmembrane protein which belongs to the class III receptor tyrosine kinase family. The extracellular region consists of five immunoglobulin-like domains where the second and third loops are thought to be involved in ligand binding (Ashman, 1997; Bühring, 1995). CD117 has been shown to be a marker for tissue mast cells, hematopoietic stem cells, and progenitor cells in normal human bone marrow. The majority of CD117-positive marrow cells (50-70%) co-express CD34 and comprise progenitor cells and their precursors of all hematopoietic lineages. Apart from mast cells, CD117 is essentially absent from mature hematopoietic cells. Although CD117 is primarily a marker for non-lymphoid progenitor cells in the hematopoietic cells/tissues, including melanocytes, embryonic brain, and the reproductive system (Ashman, 1997). Anti-CD117, 104D2, was included in the Sixth International Workshop and Conference on Human Leucocyte differentiation Antigens, and studies by different laboratories confirmed its reactivity with CD117 (Ashman, 1997).

Specificity

Antibody recognizes Human CD117 antigen.

Immunogen

MOLM-1 megakaryocytic cell line.

Application

Milli-Mark Anti-CD117 Antibody, c-kit-PE, clone 104D2 detects level of CD117 & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using TF-1 cells

Target description

145 kDa (Ashman, 1997; Buhring, 1995)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
TF-1 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service