Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.77
Input:
sample type plasma (K2 EDTA)
sample type serum
sample type serum
Assay range:
accuracy: 91-111%, linearity: 86-108%, standard curve range: 0.2-20 ng/mL
Species reactivity:
human
Storage temp.:
2-8°C
Shipped in:
wet ice
Product Name
Human C-Peptide ELISA, This Human C-Peptide ELISA is used to measure & quantify C-Peptide levels in Metabolism & Endocrine research.
Quality Segment
species reactivity
human
packaging
kit of 1 × 96 wells
parameter
10 μL sample volume (4hr assay)
assay range
accuracy: 91-111%, linearity: 86-108%, standard curve range: 0.2-20 ng/mL
technique(s)
ELISA: suitable
input
sample type plasma (K2 EDTA)
sample type serum
NCBI accession no.
UniProt accession no.
application(s)
research use
detection method
colorimetric (450nm)
shipped in
wet ice
storage temp.
2-8°C
General description
C-peptide is a byproduct of normal insulin production by the beta cells in the pancreas and correlates directly to endogenous concentrations of insulin.
Application
3 hour assay, room temperature 10 μL sample size
Research Category
Metabolism
Metabolism
Research Sub Category
Obesity
Metabolic Disorders
Obesity
Metabolic Disorders
Used to detect/quantify: C-Peptide
Disclaimer
For research use only. Not for use in diagnostic procedures.
signalword
Warning
hcodes
Hazard Classifications
Met. Corr. 1 - Skin Sens. 1
wgk
WGK 3
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

