Skip to Content
Merck

ES-002

EmbryoMax® 2X Freezing Medium for ES Cells

Formulated with 20% DMSO & Fetal Bovine Serum and available in a 10 x 10 mL format.

Synonym(s):

ES Cell Freezing Medium, EmbryoMax Freezing Medium, EmbryoMax Freezing Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EmbryoMax® 2X Freezing Medium for ES Cells, formulated with 20% DMSO & Fetal Bovine Serum, The EmbryoMax 2X Freezing Medium for ES Cells is formulated with 20% DMSO & Fetal Bovine Serum and available in a 10 x 10 mL format.

Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | stem cell: suitable, cryopreservation: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

General description

This unique formulation of cell culture freezing media is for cryopreservation of a broad spectrum of mammalian cells (see viability data). Freezing Media contain 10% of the active cryopreservation ingredient (either DMSO or Glycerol). The remaining 90% is a proprietary mixture of Calf and Fetal Bovine Serum. The reagents used in these products are certified to be virus and mycoplasma free.

The sera utilized in this medium is approved by the United States Department of Agriculture (USDA) to ensure that sera is free of diseases such as Bovine Spongiform Encephalopathy (BSE).

Packaging

10 x 10 mL

Preparation Note

Store at -80°C up to 2 years

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library