Skip to Content
Merck

EI8

Leupeptin

lyophilized powder, protease inhibitor, Chemicon®

Synonym(s):

N-Acetyl-L-leucyl-L-leucyl-L-argininal, Ac-Leu-Leu-Arg-H, Acetyl-L-leucyl-L-leucylargininal, Leupeptin hemisulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.77
eCl@ss:
32160405
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Leupeptin,

manufacturer/tradename

Chemicon®

Quality Level

shipped in

dry ice

General description

Leupeptin is a water-soluble and cell-permeable organic compound. It is produced by various species of actinomycetes and several other fungal families.

Application

Leupeptin has been used as a protease inhibitor supplement in cell lysis buffer for sample preparation.

Biochem/physiol Actions

Leupeptin serves as a lysosomal protease and calpain (serine- and cysteine-like protease) inhibitor. It may be used to reduce the cell death induced by excess calpain activation. Leupeptin confers significant protection against hair cell damage caused by gentamicin ototoxicity. In addition, it also impedes protein degradation in denervated rat muscles and muscles of mice with hereditary muscular dystrophy. Thus, leupeptin may be beneficial in hindering tissue atrophy.

Physical form

Lyophilized.

Preparation Note

Maintain dry at -20ºC for up to 18 months after date of receipt. Store reconstituted product in aliquots at -20ºC for up to 6 months. Avoid repeated thaw freeze cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Protein Blotting Survival Handbook: Tips and Tricks

Western blotting is one of the most commonly used techniques in the lab, yet difficulties persist in obtaining consistent, quality results. We’ve been helping scientists publish their Western blots for decades, with continued innovation and steadfast technical support. Explore our expanded portfolio of products, including optimized reagents for chemiluminescent and Ḁuorescent Westerns, as well as the SNAP i.d.® system, which reduces blocking, washing and antibody incubation time from hours to minutes.


P Libby et al.
Science (New York, N.Y.), 199(4328), 534-536 (1978-02-03)
The protease inhibitor leupeptin decreases protein degradation in rat skeletal and cardiac muscle incubated in vitro, while protein synthesis remains unaltered. Leupeptin also lowers protein breakdown in denervated rat muscles and affected muscles from mice with hereditary muscular dystrophy. Leupeptin
Karen Maes et al.
American journal of respiratory and critical care medicine, 175(11), 1134-1138 (2007-03-24)
Controlled mechanical ventilation (CMV) has been shown to result in elevated diaphragmatic proteolysis and atrophy together with diaphragmatic contractile dysfunction. To test whether administration of leupeptin, an inhibitor of lysosomal proteases and calpain, concomitantly with 24 hours of CMV, would
Erika Billinger et al.
FEBS open bio, 10(12), 2605-2615 (2020-10-06)
Leupeptin is a naturally occurring inhibitor of various proteases, in particular serine proteases. Following its discovery, the inhibitory properties of several other peptidyl argininals have been studied. The specificity of leupeptin is most likely due to the Leu-Leu-Argininal sequence, and



Global Trade Item Number

SKUGTIN
EI804053252741524