Skip to Content
Merck

EI10

Pepstatin A

≥90% (HPLC), lyophilized solid, aspartic proteinase inhibitor, Chemicon®

Synonym(s):

Pepstatin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.77
eCl@ss:
32160405
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Pepstatin A, Pepstatin A is an inhibitor of aspartic proteinases such as pepsin, cathepsins D and E

Quality Level

manufacturer/tradename

Chemicon®

shipped in

dry ice

General description

M.W.: 685.89

SEQUENCE: isovaleryl-L-Val-L-Val-AHMHA-L-Ala-AHMHA

Biochem/physiol Actions

Inhibitor Type: Protease Inhibitors

Physical form

Purity: >90.0%; pale yellow powder; pH (1% solution) 5.2

Preparation Note

Maintain dry at -20ºC for up to 18 months after date of receipt. Pepstatin A is soluble in either DMSO or ethanol:acetic acid (9:1) at the concentration of 1 mg/mL. Pepstatin A is insoluble in water as well as aqueous buffer at near neutral pH.

NOTE: Aqueous buffer at a slightly alkaline pH is recommended for diluting the reconstituted solution of either of the above organic solvents. If water or acetic acid is used for dilution, Pepstatin A may precipitate from the solution.

Other Notes

Replaces: EI9

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pepstatin, a new pepsin inhibitor produced by Actinomycetes.
H Umezawa et al.
The Journal of antibiotics, 23(5), 259-262 (1970-05-01)

Global Trade Item Number

SKUGTIN
EI1004053252742668

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service