Skip to Content
Merck

CC321-1

PhotoCol-LAP, Methacrylated Collagen Hydrogel Kit

Photocrosslinked type I collagen hydrogel (CC321 Kit Component 1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

cell analysis

General description

3D cell culture, including bioprinting, allows for the creation of more physiological cell models by allowing cells to simultaneously interact with integrins on all cell surfaces, resulting in the activation of specific signaling pathways not activated in traditional 2D cell culture methods. Hydrogels are water swollen polymers that allow for the culture of cells in 3-dimensions and can have profound effects on cellular development, differentiation, migration, and function. New areas of tissue engineering such as 3D bioprinting, have utilized UV photocrosslinked methacrylated hydrogel biomaterials (PEGMA, GelMA, HAMA and ColMA etc.) to encapsulate cells to make printable bioinks. The PhotoCol-LAP, Methacrylated Collagen Hydrogel Kit (CC321: CC321-1, CC321-2) is based upon purified type I bovine collagen methacrylate (ColMA), which when photocrosslinked provides a native-like 3D environment for cells. In addition to type I bovine collagen methacrylate, the kit includes the photoinitiator lithium phenyl-2,4,6-trimethylbenzoylphosphinate (LAP) and other reagents for users to easily fine tune their photocrosslinking experiments (i.e. altering hydrogel stiffness or gelling speeds). The methacrylated Type I collagen is produced from telo-peptide intact bovine collagen that has been modified by reacting with the protein′s free amines, primarily the ɛ-amines groups of the lysine residues as well as the α-amines groups on the N-termini. >20% of the total lysine residues of the collagen molecule have been methacrylated. The collagen is extracted from bovine hide and contains a high monomer content that was isolated from a closed herd and purified using controlled manufacturing processes.PhotoCol is a registered trademark of Advanced BioMatrix, Inc.

Application

Photocrosslinking is initiated using a 405 nm blue light source.

Packaging

1 Kit

Analysis Note

Sterility (USP modified): No GrowthEndotoxin LAL: ≤ 10.0 EU/mlGel Formation: ≤ 40 minKinetic Gel Test: ≤ 40 minPurity (SDS Page Silver Stain): ≥ 99%Degree of Methacrylation: ≥ 20%pH: 3.0-4.0Osmolality: ≤ 40 mOsm/Kg H20Young′s Modulus: CharacteristicElectrophoretic Pattern: Characteristic

Other Notes

CC321-1 (Store at Room Temp): Neutralization Solution, 1 X 10 mL (CS226411).
Note: The complete PhotoCol-LAP, Methacrylated Collagen Hydrogel Kit (CC321) consists of CC321-1 and CC321-2 sold seperately.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library