Skip to Content
Merck

CBL561

Anti-CD24 Antibody, clone SN3

clone SN3, Chemicon®, from mouse

Synonym(s):

Heat Stable Antigen, HAS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
SN3, monoclonal
Application:
FACS, IHC, IP
Citations:
10
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

SN3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD24(100133941)

Immunogen

Glycoproteins purified from human NALM-1 cell line.

Application

Anti-CD24 Antibody, clone SN3 detects level of CD24 & has been published & validated for use in FC, IP, IH.
Immunocytochemistry on frozen tissue sections.

Flow Cytometry: 1:50 - 1:100 per 10e6 cells in 100 μL final volume.

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Biochem/physiol Actions

The antibody reacts with the CD24 antigen a heavily glycosylated molecule that migrates as a broad band of 35-45 kDa on both reducing and non-reducing SDS gel electrophoresis. CD24 is attached to the cell membrane via a glycosyl phosphatidylinositol (GPI) anchor and is expressed at multiple stages of B cell development, beginning with the bone marrow CD34 positive pro-B cell compartment and continuing through mature surface IgM positive/IgD positive B cells. CD24 is also expressed on the vast majority of B-lineage acute lymphoblastic leukaemias, B cell chronic lymphocytic leukaemias and B cell non-Hodgkin′s lymphoma. In the normal spleen, mantle zone lymphocytes are CD24 strongly positive, but germinal center cells are CD24 negative or CD24 weakly positive.

Physical form

Affinity purified immunoglobulin by Protein G chromatography. Liquid in 1 mL of PBS, pH 7.4 containing no preservatives.
Format: Purified

Preparation Note

Maintain at -20°C for up to 12 months from date of receipt in undiluted aliquots.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library