Skip to Content
Merck

CBL162

Anti-CD9 Antibody, clone MM2/57

clone MM2/57, Chemicon®, from mouse

Synonym(s):

MRP-1, DRAP-27

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
MM2/57, monoclonal
Application:
FACS, IHC, IP, WB
Citations:
41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MM2/57, monoclonal

species reactivity

human, rabbit, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD9(928)

Related Categories

Immunogen

Human platelet glycoprotein

Application

Anti-CD9 Antibody, clone MM2/57 detects level of CD9 & has been published & validated for use in FC, IP, WB, IH.
Immunohistochemistry: Frozen sections of platelets, monocytes and pre-B cells with differential staining on lymphoid and epithelial tissues
Platelet inhibition and aggregation studies
Optimal working dilutions must be determined by the end user. the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Biochem/physiol Actions

This antibody reacts with a 25 kDa molecular weight band on gels of non-reduced platelet membranes and immunoprecipitates a similar band from Iodine125-labeled platelets. The antigen is expressed on eosinophils, granulocytes, monocytes and numerous stromal cells. This CD9 antigen belongs to the TM4 superfamily of cell surface proteins with the N- and C-terminals localized in the cytoplasm.

Physical form

Format: Purified
Protein A purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Preparation Note

1 year at 4°C from date of shipment

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry Ter-Ovanesyan et al.
eLife, 10 (2021-11-17)
Extracellular vesicles (EVs) are released by all cells into biofluids and hold great promise as reservoirs of disease biomarkers. One of the main challenges in studying EVs is a lack of methods to quantify EVs that are sensitive enough and
V Zvereff et al.
British journal of cancer, 97(7), 941-948 (2007-09-13)
CD9, a member of the tetraspanin family of proteins, is involved in a variety of cellular interactions with many other proteins and molecules. Although CD9 has been implicated in cell fusion, migration and cancer progression, the detailed function of this
Yuan-Yin Chen et al.
Cancers, 13(21) (2021-11-14)
C-Circles, self-primed telomeric C-strand templates for rolling circle amplification, are the only known alternative-lengthening-of-telomeres (ALT)-specific molecule. However, little is known about the biology of C-Circles and if they may be clinically useful. Here we show that C-Circles are secreted by
Xiaoman Zhang et al.
PloS one, 17(6), e0270634-e0270634 (2022-06-25)
Extracellular vesicles (EVs) have attracted much attention as potential diagnostic biomarkers for human diseases. Although both plasma and serum are utilized as a source of blood EVs, it remains unclear whether, how and to what extent the choice of plasma
Yesim Verel-Yilmaz et al.
Frontiers in cell and developmental biology, 9, 697939-697939 (2021-08-10)
Due to a grim prognosis, there is an urgent need to detect pancreatic ductal adenocarcinoma (PDAC) prior to metastasis. However, reliable diagnostic imaging methods or biomarkers for PDAC or its precursor lesions are still scarce. ADAM8, a metalloprotease-disintegrin, is highly

Related Content

As the focus of stem cell research undergoes a transition from animal to human models, researchers are in critical need of validated products to support the isolation, maintenance, differentiation, and characterization of human stem cells. While many reagents designed for rodent systems can be applied to human stem cell studies, they are not truly optimized for robust human stem cell culture or analysis. This is why human stem cell researchers have always trusted EMD Millipore, the first provider of commercially available human embryonic stem cells and human neural stem cell lines, to accelerate their research. All of our human stem cell systems are extensively tested in defined media culture, and differentiated progeny are comprehensively characterized with highly validated antibodies and detection reagents.

Global Trade Item Number

SKUGTIN
CBL16204053252320408

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service