Skip to Content
Merck

CBA098

InnoCyte Monocyte Cell Migration Assay

Synonym(s):

Monocyte Migration Assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352207
Usage:
sufficient for 96 tests
Storage temp.:
no temp limit
Detection method:
fluorometric
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 96 tests

Quality Segment

packaging

pkg of 1 96-well plate(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

input

sample type monocyte(s)

detection method

fluorometric

shipped in

wet ice

storage temp.

no temp limit

General description

The InnoCyte Monoctye Cell Migration Assay is designed to study the effects of various drugs or agents on monocyte motility or for identifying chemoattractant molecules. The assay is not intended for adherent cell lines, such as fibroblasts or solid tumor cell lines.

Biochem/physiol Actions

The Calbiochem InnoCyte 96-Well Monoctye Cell Migration Assay is intended for the study of the effects of various drugs or agents on monocyte motility or for identifying chemoattractant molecules. The assay is not intended for adherent cell lines, such as fibroblasts or solid tumor cell lines. To study these cell types, please use Cat. No. CBA010 or CBA017.

Preparation Note

Cell Labeling Mixture: Warm the D-PBS and Calcein-AM Solution to room temperature. Add 35 µl Calcein-AM Solution to 10 ml D-PBS. Mix well. Note: Prepare just before use (see step 7 of the Detailed Protocol below).
Upon arrival store the Calcein-AM at -20°C and the remaining components at 4°C.

Other Notes

96-Well Monocyte Cell Migration Chamber, Black, Conical 96-Well Plate, D-PBS, Calcein-AM, and a user protocol.
Cravens, P.D. 2007. Scan. J. Immunol.65, 514.
Charo, I.F., et al. 2006. N. Engl. J. Med.354, 610.
Knitscher, U., et al. 2000. Eur. J. Pharmacol.401, 259.
Turner, L., et al. 1999. Eur. J. Immunol.29, 2288.
Cross, A.K., et al. 1997. Cytokine9, 521.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

10-13 - German Storage Class 10 to 13

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library