Skip to Content
Merck

APT173

Caspase 9 Colorimetric Activity Assay Kit, LEHD

Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.

Synonym(s):

Mch-6, ICE-LAP6, Apaf-3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

detection method

colorimetric

shipped in

dry ice

Gene Information

human ... CASP9(842)

General description

Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. Chemicon′s Caspase-9 Colorimetric Activity Assay Kits provide a simple and convenient means for assaying the activity of caspases that recognize the LEHD. The assay is based on spectophotometric detection of the chromophore p-nitroaniline (pNA) after cleavage from the labeled substrate LEHD-pNA. The free pNA can be quantified using a spectrophotometer or a microtiter plate reader at 405 nm. Comparison of the absorbance of pNA from an apoptotic sample with an uninduced control allows determination of the fold increase in caspase-9 activity.

Application

Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.
Research Category
Apoptosis & Cancer

Preparation Note

Store kit materials at -20°C up to their expiration date.

Special Precautions:

· After thawing reagents, use immediately or aliquot and freeze at -20°C for longer storage. Avoid repeated freeze/thaw cycles.

· The Caspase-9 substrate and pNA standard are especially light sensitive. Maintain these reagents in amber or covered containers.

Other Notes

5X Cell Lysis Buffer (Part No. 90065): 5 mL

5X Assay Buffer (Part No. 90066): 10 mL

Caspase-9 Substrate (Ac-LEHD-pNA) (Part No. 90084): 1 mL of 3 mg/mL solution

Caspase-9 Inhibitor (Ac-LEHD-CHO) (Part No. 90087): 50 μL of 100 μM (0.054 mg/mL) in DMSO

pNA Standard (Part No. 90085): 250 μL of 10 mM in DMSO

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Dimosthenis Mantopoulos et al.
PloS one, 6(9), e24245-e24245 (2011-10-01)
Detachment of photoreceptors from the underlying retinal pigment epithelium is seen in various retinal disorders such as retinal detachment and age-related macular degeneration and leads to loss of photoreceptors and vision. Pharmacologic inhibition of photoreceptor cell death may prevent this
Jean F Soustiel et al.
Experimental neurology, 214(2), 201-208 (2008-09-16)
The 18 kDa translocator protein (TSPO) is a protein complex located at the outer mitochondrial membrane and interacting with the mitochondrial permeability transition pore (mPTP), indicating its involvement in the control of mPTP opening. We intended to explore the effect
Mi In Roh et al.
Investigative ophthalmology & visual science, 52(6), 3825-3831 (2011-02-12)
To investigate whether edaravone (3-methyl-1-phenyl-2-pyrazolin-5-one), a free radical scavenger, would be neuroprotective against photoreceptor cell death in a rat model of retinal detachment (RD). RD was induced in adult Brown Norway rats by subretinal injection of sodium hyaluronate. Edaravone (3



Global Trade Item Number

SKUGTIN
APT17308436037123344