AP194C
Donkey Anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed
Chemicon®, from donkey
Synonym(s):
Chicken IgG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
donkey
Quality Level
conjugate
CY3 conjugate
antibody form
F(ab′)2 fragment of affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
chicken
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
Application
Detect Donkey Chicken IgY using this Donkey anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed validated for use in IF.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of clinical investigation, 120(9), 3191-3208 (2010-08-04)
Human cytomegalovirus (HCMV) in clinical material cannot replicate efficiently in vitro until it has adapted by mutation. Consequently, wild-type HCMV differ fundamentally from the passaged strains used for research. To generate a genetically intact source of HCMV, we cloned strain
Non-epithelial stem cells and cortical interneuron production in the human ganglionic eminences.
Nature Neuroscience null
NeuroImage, 256, 119277-119277 (2022-05-07)
Biophysical models of diffusion in white matter have been center-stage over the past two decades and are essentially based on what is now commonly referred to as the "Standard Model" (SM) of non-exchanging anisotropic compartments with Gaussian diffusion. In this
Brain : a journal of neurology, 140(6), 1692-1705 (2017-04-27)
A biomarker that will enable the identification of patients at high-risk for developing post-injury epilepsy is critically required. Microvascular pathology and related blood-brain barrier dysfunction and neuroinflammation were shown to be associated with epileptogenesis after injury. Here we used prospective
Biomolecules, 12(7) (2022-07-28)
The pathological events of age-related macular degeneration are characterized by degenerative processes involving the photoreceptor cells, retinal pigment epithelium (RPE), and the Bruch's membrane as well as choroidal alterations. To mimic in vivo interactions between photoreceptor cells and RPE cells
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service