Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
-
biological source
goat
Quality Segment
conjugate
FITC conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
concentration
2.0 mg/mL
technique(s)
immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
Application
Research Category
Secondary & Control Antibodies
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
Fragment Specific Secondary Antibodies
This Goat anti-Human IgM Antibody, Fc5μ, FITC conjugate is validated for use in IF for the detection of Human IgM.
Biochem/physiol Actions
Based on immunoelectrophoresis, the antibody reacts with the Fc5μ portion of the human IgM heavy chain. No antibody was detected against normal human IgG or IgA, or against non-immunoglobulin serum proteins, but the antibody may cross-react with immunoglobulins from other species.
Physical form
PRESENTATION: Liquid. Buffer = 0.02 M Phosphate Buffered Saline, 0.25 M NaCl, 0.1% NaN3, 15 mg Bovine Serum Albumin per mL of buffer for stabilizer.
F/P RATIO:
6.0 micrograms FITC per mg of antibody.
F/P RATIO:
6.0 micrograms FITC per mg of antibody.
Preparation Note
Store in the dark at +2-8°C. Do not freeze.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AP114F | 08436037125584 |