Skip to Content
Merck

AP112A

Goat Anti-Human IgG Antibody, Alkaline Phosphatase conjugate

Chemicon®, from goat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
alkaline phosphatase conjugate
Clone:
polyclonal
Application:
ELISA, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Alkaline Phosphatase-conjugated Affinity Purified Goat anti-Human IgG (H+L)

Application

Detect Human IgG using this Goat anti-Human IgG Antibody, Alkaline Phosphatase conjugate validated for use in ELISA & WB.
EIA and Western blots: 1:5,000-1:50,000.

Immunohistochemistry: 1:1,000-1:2,000.

Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Biochem/physiol Actions

Based on immunoelectrophoresis, the antibody reacts with the heavy chains on human IgG and with light chains common to most human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins, but the antibody may cross-react with immunoglobulins from other species.

Physical form

Lyophilized. Buffer = 0.01 M Tirs HCl, 0.25 M NaCl, pH 8.0 with

15 mg/mL BSA and 0.05% sodium azide.

RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature.
Serum

Preparation Note

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. After dilution do not use for more than one day. For extended storage after reconstitution, add an equal volume of glycerol (ACS grade for better) to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

WARNING:

For research use only; not for use as a diagnostic.

Other Notes

Replaces: MABN1055

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
AP112A04053252587771