Skip to Content
Merck

AP101F

Rabbit Anti-Human IgG Antibody, FITC conjugate

Chemicon®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

DILUTION:

Suggested dilution for most applications: 1:50-1:200

Optimal working dilutions must be determined by the end user.
This Rabbit anti-Human IgG Antibody, FITC conjugate is validated for use in IF for the detection of Human IgG.

Biochem/physiol Actions

Human IgG (H&L)

FLUOROPHORE/PROTEIN:

Approximately 6.9 mg/mg; 2.6 moles FITC per mol IgG

Physical form

Liquid in 0.01 M Phosphate buffer, 0.25 M NaCl pH 7.6 with 15 mg/mL BSA and 0.05% sodium azide.

RECONSTITUTION:

Reconstitute to 1 mg/mL with sterile distilled water.

Preparation Note

Maintain lyophilized product between 2 and 8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


hcodes

Hazard Classifications

Aquatic Chronic 3

supp_hazards

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
AP101F04053252405235